Orders LinkContactUsLinkComponent
Ice borers
Ice borers
Supplier:   BURKLE
Ice borers
Supplier:  BURKLE
Certificates
Ice borers
3 Options Available Below
Product Details & Documents
Sampler for deep-frozen products in the food and luxury items sector, and for semi-solid substances. The very robust hollow drill cuts quickly into the sample material with its sharp sawing crown. A sample cylinder (cross-section sample) can be cut out.
  • Stainless steel AISI 316L (1.4404) / AISI 316 Ti (1.4571)
  • Sterilisable
  • Cross-section sample
The ice drill can be mounted on a standard commercial drilling machine. The core bit drills quickly and easily down to the desired depth in the frozen material. This makes it possible to cut a representative core sample from all layers of the frozen medium without expending any great effort. Drilling core samples can be taken from a drilling depth of up to 100 or 200 mm, depending on the design. The ejector makes it easy to release the sample from the drill head.
The drill is sterilisable and suitable for use with foods. The ice drill is ideal for quality control in the food and animal feed industries, and is also suitable for sampling frozen meat for meat inspections. Other deep-frozen substances can be sampled easily as well with the sampling drill, such as frozen liquids.
Delivery information: Complete with case.
Documents
Product Family Options
Product Information
  • Immersion depth
    Ø
    Availability
    Price
  • 300-0357
    Ice borer
    Ice borer
    100 mm
    Inner/outer: 8/10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 212,00 €
    212,00 €
    1 * 1 ST
       212,00 €
  • 312-0072
    Ice borer
    Ice borer
    200 mm
    Inner/outer: 16/21 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 538,00 €
    538,00 €
    1 * 1 ST
       538,00 €
  • 312-0086
    Ice borer
    Ice borer
    200 mm
    Inner/outer: 50/54 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 936,00 €
    936,00 €
    1 * 1 ST
       936,00 €