Conical hand drill, QualiRod
Supplier: BURKLE
Jump to:
About this item
Conical hand drill with sharp cutting edges and a solid handle made from stainless steel V2A (1.4301)/AISI 304.
- Sterilisable
Designed to cut out samples from soft, paste-like substances (cheese, butter, clay, loam, soap and wax).
Certificeringen: ISO 707
2 Options Available Below
Product Details & Documents
Conical hand drill with sharp cutting edges and a solid handle made from stainless steel V2A (1.4301)/AISI 304.
- Sterilisable
Designed to cut out samples from soft, paste-like substances (cheese, butter, clay, loam, soap and wax).
Certificeringen: ISO 707