ContactUsLinkComponent
Hygiene monitoring system, HY-LiTE® 2
Hygiene monitoring system, HY-LiTE® 2
Catalog # 1.30100.0001
Supplier:  Merck
Hygiene monitoring system, HY-LiTE® 2
Catalog # 1.30100.0001
Supplier:  Merck
Specifications
  • Range:
    Linear: 0 – 99.000 RLU, Logarithmic: 0 – 5.00 log 10 RLU
  • Description:
    HY-LiTE® 2 system
  • W×H×D:
    11×13×28 cm
  • Connections:
    Two serial RS232 interfaces for data transfer between system and a computer; One low-voltage socket for the HY-LiTE® power supply unit; One ROM key port
  • Pk:
    1
Specifications
About this item
Rapid, quantitative hygiene monitoring in liquids and on surfaces with ATP-bioluminescence HY-LiTE® 2, contained in a convenient lightweight shoulder bag with data logging and TREND2 software for advanced data management. The HY-LiTE® TREND2 software can be used to compile HACCP plans and individual limits. Graphs showing results make them easy to evaluate and interpret. HY-LiTE® is already used in a wide variety of production and processing fields, like the food industry, packing industry, services (cleaning of buildings), community catering and large scale kitchens, drinks manufacturers, state-owned institutions or the pharmaceutical industry.
  • Simple to use and easily portable, can be used anywhere by anyone
  • Produces results within one minute: Can be used by production workers immediately after cleaning, allowing any corrective action to be taken before production begins
  • Luminometer holds data from 2000 tests enabling trends to be identified as well as seeing the 'before and after' effect of cleaning procedures
  • Wide operating temperature range (5 to 35 °C), can be used anywhere from cold stores to hot food processing areas
Delivery: Includes HY-LiTE® 2 luminometer, quality assurance certificate and test report, A/C power supply (110 - 240 V) and adapters for international use, battery holder (requires 4 x AA batteries; batteries not included), USB stick with TREND 2 software for Windows O/S PCs (Win10 supported) and user manuals, USB cable for TREND 2 data synchronization, and robust carrying case with shoulder strap. In addition, several additional HY-LiTE® 2 accessories are also available to order as separate items; contact your Avantor representative for more information.