Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sampling pot, Liquid CupSampler

Sampling pot, Liquid CupSampler

Sampling pot, Liquid CupSampler, Stainless steel, 100 ml, 1000 mm, 55 mm
Catalog # 300-0025
Supplier:   BURKLE
Sampling pot, Liquid CupSampler
Catalog # 300-0025
Supplier:  BURKLE
Sampling pot, Liquid CupSampler
Sampling pot, Liquid CupSampler

Sampling pot, Liquid CupSampler, Stainless steel, 100 ml, 1000 mm, 55 mm

Price
1 280,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Liquid CupSampler collects liquids of varying viscosity. Media containing particles can also be sampled without difficulty.

  • Cup can be unscrewed from the stem for cleaning

The sampler can be used in chemistry, foodstuffs, and cosmetics.

Specifications
  • Ø:
    55 mm
  • Capacity:
    100 ml
  • Description:
    Sampling pot, Liquid CupSampler
  • Length:
    1000 mm
  • Material:
    Stainless steel