Order Entry
Belgium
ContactUsLinkComponent
 
Nucleic acid and protein purification systems, KingFisher™
  735-0273
undefined
Nucleic acid and protein purification systems, KingFisher™
  735-0273

 

  • Plate types:
    Microstrips 1×8 wells, 8×12 grid format, 20 - 100 and 20 - 200 μl well volumes
  • Description:
    Purification system KingFisher™
  • Seq:
    01
  • Model:
    KingFisher™
  • Incubation:
    Ambient, no heating
  • Interfaces:
    Serial RS232C port
  • Int. software:
    Space for ~ 8 internal protocols
  • Keyboard/display:
    START/STOP/two cursor keys/LCD
  • Magnet rods:
    2×12 format
  • Processing volume:
    20 - 200 µl
  • Robot compatibility:
    No
  • Sample vessel (disposable):
    8×12 strip plate (100 µl or 200 µl), capacity for 2 plates
  • Tip combs (disposable):
    Special design, 1×12 format
  • Weight:
    10 kg
  • W×D×H:
    290×290×310 mm
  • Capacity (samples/run):
    24
  • Magnetic particle size:
    ~ >1 μm
  • Pk:
    1

 

 

The KingFisher™ magnetic particle processors are designed to automate time consuming sample processing of nucleic acids, proteins or cells from virtually any source (blood, cell cultures, tissue lysates, soil, faeces, for example). The KingFisher™ family consists of instruments with different throughput and volume capacities to fulfil individual sample processing needs. Each system consists of an instrument, specially designed plastics, and software. The KingFisher™ systems offer rapid and reproducible purification of high quality DNA, RNA, proteins and cells for various types of downstream applications, including genomics and proteomics, drug discovery, forensics, biomarker discovery, quality control, and veterinary assays.

  • High speed, fully automated purification
  • Purification of any sample from any source
  • Ready made and custom protocols
  • Easy operation via instrument or external PC

KingFisher™ has the ability to purify small scale samples economically. The system can run up to 24 samples in 20 to 200 μl. All purification and processing steps are carried out in microstrips, with simple push button operation.