Orders LinkContactUsLinkComponent
Single channel pipettes, electronic, variable volume, Ovation®
Single channel pipettes, electronic, variable volume, Ovation®
Certificates
Single channel pipettes, electronic, variable volume, Ovation®
Single channel pipettes, electronic, variable volume, Ovation®
4 Options Available Below
Product Details & Documents
Manually operated pipette with quick set digital volume adjustment.
  • Easy to program and calibrate for different environmental conditions and liquids
  • Pipettes stand flat on the bench so no pipette stands are required
  • Motor-driven volume adjustment for ease of change
  • One-way disposable nozzle with built-in filter to prevent pipette contamination
  • Colour-coded for easy volume recognition
* Certified DNase/RNase free
 : Supplied with a lithium ion battery (giving approximately six months use).
Product Family Options
Ovation electronic pipettes
Product Information
  • Volume
    Increment
    Accuracy
    Imprecision
    Colour code
    Packed
    Availability
    Price
  • 2 - 20 µl
    0,02 µl
    ±7 - ±1 %
    <2 - <0,3 %
    Yellow
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 844,00 €
    844,00 €
    1 * 1 ST
       844,00 €
  • 10 - 100 µl
    0,1 µl
    ±3,5 - ±0,8 %
    <1 - <0,15 %
    Green
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 844,00 €
    844,00 €
    1 * 1 ST
       844,00 €
  • 20 - 200 µl
    0,2 µl
    ±2,5 - ±0,8 %
    <1 - <0,15 %
    Blue
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 844,00 €
    844,00 €
    1 * 1 ST
       844,00 €
  • 100 - 1000 µl
    1 µl
    ±3 - ±0,8 %
    <0,6 - <0,15 %
    Purple
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 070,00 €
    1 070,00 €
    1 * 1 ST
       1 070,00 €
Ovation pipette tips
Product Information
  • Catalog #
    Sterile
    Volume
    Tip style
    Packed
    Availability
    Price
  • -
    1400 µl
    Standard
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 95,70 €
    95,70 €
    1 * 1.000 ST
       95,70 €
Product Information
  • Catalog #
    Sterile
    Volume
    Tip style
    Packed
    Availability
    Price
  • +
    250 µl
    VistaRak™, pyrogen-free, RNase/DNase certified
    5 racks, 192 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 213,00 €
    213,00 €
    1 * 960 ST
       213,00 €
Product Information
  • Catalog #
    Sterile
    Volume
    Tip style
    Packed
    Availability
    Price
  • +
    250 µl
    Pyrogen-free, RNase/DNase certified
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 0,00 €
    0,00 €
    1 * 960 ST
       0,00 €
Product Information
  • Catalog #
    Sterile
    Volume
    Tip style
    Packed
    Availability
    Price
  • -
    25 µl
    Standard
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 70,60 €
    70,60 €
    1 * 1.000 ST
       70,60 €
Product Information
  • Catalog #
    Sterile
    Volume
    Tip style
    Packed
    Availability
    Price
  • -
    35 µl
    Standard
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 ST
    1 * 960 ST - 107,00 €
    107,00 €
    1 * 960 ST
       107,00 €
Product Information
  • Catalog #
    Sterile
    Volume
    Tip style
    Packed
    Availability
    Price
  • -
    1250 µl
    VistaStak™
    3 rack layers, 192 each
    Log in for availability
    Log in for availability
    1 * 576 ST
    1 * 576 ST - 79,00 €
    79,00 €
    1 * 576 ST
       79,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • VITON seal kit for Ovation 20 - 200 µl (QS), 101 - 200 µl (F1 and F2)
    VITON seal kit for Ovation 20 - 200 µl (QS), 101 - 200 µl (F1 and F2)
    VIST9057-2035
    Log in for availability
    1 * 1 KIT - 52,70 €
Product Information
  • Catalog #
    Availability
    Price
  • VITON seal kit for Ovation 100 - 1000 µl (QS), 201 - 1000 µl (F1 and F2)
    VITON seal kit for Ovation 100 - 1000 µl (QS), 201 - 1000 µl (F1 and F2)
    VIST9057-3036
    Log in for availability
    1 * 1 KIT - 52,70 €