Orders LinkContactUsLinkComponent
Skin section
Skin section
Skin section, 240×150×35 mm
Catalog # 761-0007
Marketsource
Supplier:   3B Scientific
Skin section
Catalog # 761-0007
Supplier:  3B Scientific
Marketsource
Skin section
Skin section
Skin section, 240×150×35 mm
Price
79,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The two halves of this relief model show the three layers of hairy and hairless skin in order to make the differences clear.
In detail with hair follicles, sebaceous glands, sweat glands, receptor, nerves and vessels.
Ordering information: On base.
Specifications
  • Description:
    Skin section
  • W×D×H:
    240×150×35 mm
  • Magnification:
    40×