Order Entry
Belgium
Orders LinkContactUsLinkComponent
Eye, 3×

Eye, 3×

Eye in six parts, 90×90×150 mm
Catalog # 761-0005
Marketsource
Supplier:   3B Scientific
Eye, 3×
Catalog # 761-0005
Supplier:  3B Scientific
Marketsource
Eye, 3×
Eye, 3×

Eye in six parts, 90×90×150 mm

Price
99,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This model dissects into both halves of sclera with cornea and eye muscle attachments, both halves of the choroid with iris and retina, lens and vitreous humour.

  • On a base
  • Six parts
  • Three times full-size

Dimensions: 9×9×15 cm

Specifications
  • Description:
    Eye in six parts
  • W×D×H:
    90×90×150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.