Orders LinkContactUsLinkComponent
Accessories for analytical Mill, A 11 basic
Accessories for analytical Mill, A 11 basic
Supplier:   IKA
Accessories for analytical Mill, A 11 basic
Supplier:  IKA
Certificates
Accessories for analytical Mill, A 11 basic
Accessories for analytical Mill, A 11 basic
7 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • For
    Availability
    Price
  • 412-0001
    Spare 80 ml grinding chamber
    Spare 80 ml grinding chamber
    Low temperature applications (-200 °C). ETFE and reinforced glass-fibre with stainless steel inlet, has good chemical resistance
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 545,00 €
    545,00 €
    1 * 1 ST
       545,00 €
  • 412-1112
    250 ml grinding chamber
    250 ml grinding chamber
    Use with double beater 431-2113. Polycarbonate with stainless steel inlet. Not suitable for Use with liquid nitrogen
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 738,00 €
    738,00 €
    1 * 1 ST
       738,00 €
  • 412-1101
    Cutting blade A11.2
    Cutting blade A11.2
    Pulverising soft fibrous materials
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 220,00 €
    220,00 €
    1 * 1 ST
       220,00 €
  • 412-1102
    Beater A11.3
    Beater A11.3
    Pulverising substances with a Mohs hardness of up to 9
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 274,00 €
    274,00 €
    1 * 1 ST
       274,00 €
  • 412-1100
    Spare beater A11.1
    Spare beater A11.1
    Pulverising substances with a Mohs hardness of up to 6
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €
  • 431-2113
    Double beater
    Double beater
    Use up to Mohs hardness 3. Can only be Used with 250 ml grinding chamber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 586,00 €
    586,00 €
    1 * 1 ST
       586,00 €
  • 431-2114
    Funnel, A 11.7
    Funnel, A 11.7
    Use with liquid nitrogen, prevents splashing when pouring
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 301,00 €
    301,00 €
    1 * 1 ST
       301,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.