Order Entry
Belgium
ContactUsLinkComponent
Polarising microscope, PM-1805
Polarising microscope, PM-1805
Supplier:  MOTIC
Certificates
undefined
Polarising microscope, PM-1805
Supplier:  MOTIC

Specifications

  • Catalog No:
  • 630-1469
  • 630-1169
  • Head:
  • 45° inclined, 360° rotating
  • Eyepieces:
  • 10×/18 mm with pointer adjustment
  • Nosepiece:
  • Quadruple
  • Objectives:
  • Achromatic 4×, 10×, 40× (spring)
  • Stage:
  • Ø 130 mm
  • Condenser:
  • Abbe 1,25 N.A. with iris diaphragm and filter holder
  • Illumination:
  • Halogen

Specifications

Related Information

Eyepieces

Product Information

Frequently Bought Together