Orders LinkContactUsLinkComponent
Binocular magnifiers
Binocular magnifiers
Supplier:   ESCHENBACH OPTIK
Binocular magnifiers
Supplier:  ESCHENBACH OPTIK
Certificates
Binocular magnifiers
Binocular magnifiers
3 Options Available Below
Product Details & Documents
Magnifiers with discrete metal frame and black matt finish for use with monocular lens components.
* Shorter working distance due to integrated convergence supporting prisms.
Delivery information: Supplied with a protective box.
Caution: Not suitable for spectacle wearers.
Product Family Options
Product Information
  • Magnification
    Availability
    Price
  • 630-8214
    Binocular magnifying lens with 250 mm working distance*
    Binocular magnifying lens with 250 mm working distance*
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 191,00 €
    191,00 €
    1 * 1 ST
       191,00 €
  • 630-8215
    Binocular magnifying lens with 180 mm working distance
    Binocular magnifying lens with 180 mm working distance
    2,5×
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 191,00 €
    191,00 €
    1 * 1 ST
       191,00 €
  • 630-8216
    Binocular magnifying lens with 130 mm working distance
    Binocular magnifying lens with 130 mm working distance
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 191,00 €
    191,00 €
    1 * 1 ST
       191,00 €