Orders LinkContactUsLinkComponent
Reading magnifiers
Reading magnifiers
Reading magnifiers
Reading magnifiers
This magnifier provides biconvex PXM® lightweight lens for an ergonomic use and optimised performances.
  • Additional plano-convex lens in the handle for 5× magnification
  • Eyelet in the handle, round-the-neck carry cord supplied
  • Antistatic coating
Product Family Options
  • Description
    Magnification
    Ø
  • Magnifier, hand-held
    100 mm
    1 * 1 ST
    1 * 1 ST - 93,10 €
    93,10 €
    1 * 1 ST
       93,10 €
  • Magnifier, hand-held
    65 mm
    1 * 1 ST
    1 * 1 ST - 59,70 €
    59,70 €
    1 * 1 ST
       59,70 €