UV inspection lamp, B-100AP
Fournisseur: UVP ULTRA VIOLET PRODUCTS
Certificats
À propos de cet article
This lamp provides 100 W of high intensity, longwave (365 nm) illumination. It can be used for quality control, industrial inspection, and non-destructive testing.
- Lamp head (Wר: 248×152 mm) rotates 360° when attached to base, for hands free operation
- Cool-Touch™ housing keeps lamp cool to the touch
- Filter is recessed to prevent breakage
Informations de commande: Includes. 100 W spot bulb, transformer, and lead. Replacement bulbs sold separately.
Frequently Bought Together