Automatic gas cylinder change-over unit
Catalog # WSTGPC-30
Supplier: WEST GROUP
Specifications
- Description:Automatic gas cylinder change-over unit
- Pk:1
Specifications
About this item
For incubator gases CO₂, nitrogen, air.
- Manometer 3 bar
- With alarm by change-over