Dispersing elements for homogenisers
Type N: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by all methods, max. temperature 180 °C.
Type NK: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by chemical methods, max. temperature 120 °C.
Type KV: Materials in contact with medium are FFPM/SIC and stainless steel AISI 316L, suitable for vacuum down to 1 mbar, pressure up to 6 bar and for solvents, can be sterilised by chemical methods, max. temperature 220 °C.
SMK jet mixer head: The vertical flow and the high circumferential speed up to 10000 rpm ensure intensive mixing. The head is used for adding gases or liquids, for lump-free suspension of difficult to dissolve powders or for dissolving sedimented, already hardened substances.
Type KD: Materials in contact with the medium are PTFE, FFKM O-ring and stainless steel AISL 316L, suitable for solvents, can be sterilised by all methods, max. temperature 150 °C.
Caution: More dispersing elements are available on request. Please contact Avantor for details.
ST= sawtooth.
Type N: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by all methods, max. temperature 180 °C.
Type NK: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by chemical methods, max. temperature 120 °C.
Type KV: Materials in contact with medium are FFPM/SIC and stainless steel AISI 316L, suitable for vacuum down to 1 mbar, pressure up to 6 bar and for solvents, can be sterilised by chemical methods, max. temperature 220 °C.
SMK jet mixer head: The vertical flow and the high circumferential speed up to 10000 rpm ensure intensive mixing. The head is used for adding gases or liquids, for lump-free suspension of difficult to dissolve powders or for dissolving sedimented, already hardened substances.
Type KD: Materials in contact with the medium are PTFE, FFKM O-ring and stainless steel AISL 316L, suitable for solvents, can be sterilised by all methods, max. temperature 150 °C.
Caution: More dispersing elements are available on request. Please contact Avantor for details.
ST= sawtooth.
- Catalog No:
- 442-0300
- 442-0310
- 431-0251
- 442-0312
- 431-0252
- 431-2602
- 431-2603
- 431-0311
- IKAA20002973
- IKAA20002975
- 431-0312
- 431-0313
- 431-2455
- 431-2461
- 431-2458
- 431-2451
- 431-0253
- 431-2452
- 431-0254
- 431-2453
- 431-0222
- 431-2459
- 431-2456
- 431-0223
- 431-0249
- 431-0257
- 431-0255
- 431-0256
- 431-2569
- 431-2560
- 431-0258
- 431-2563
- 431-2581
- 431-2582
- Immersion depth:
- 27 - 85 mm
- Ultimate fineness:
- Suspension: 10 - 50 µm; Emulsion: 1 - 10 µm
- Materials in contact with medium:
- PTFE, AISI 316L
- Max. temperature:
- 180 °C
Product Family Options
- DescriptionTypeForSpeedStator/rotor ØLengthVolume
- S 10 N - 5 GT 10 basic6 m/s5/3,8 mm92 mm0,5 - 10 ml1 * 1 ST1 560,00 €
Specifications
Cat. No.442-0300Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 10 N - 8 GT 10 basic9,6 m/s8/6,1 mm115 mm1 - 50 ml1 * 1 ST1 560,00 €
Specifications
Cat. No.442-0310Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 10 N - 8 G - STT 10 basic9,6 m/s8,0/6,1 mm115 mm1 - 50 ml1 * 1 ST1 690,00 €
Specifications
Cat. No.431-0251Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 10 N - 10 GT 10 basic11,9 m/s10/7,6 mm115 mm1 - 100 ml1 * 1 ST1 560,00 €
Specifications
Cat. No.442-0312Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 10 N - 10 G - STT 10 basic11,9 m/s10,0/7,6 mm115 mm1 - 100 ml1 * 1 ST1 690,00 €
Specifications
Cat. No.431-0252Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 18 N - 10 GT 18 basic9,4 m/s10,0/7,5 mm108 mm1 - 100 ml1 * 1 ST1 320,00 €
Specifications
Cat. No.431-2602Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 18 N - 19 GT 18 basic15,9 m/s19,0/12,7 mm204 mm10 - 1500 ml1 * 1 ST1 350,00 €
Specifications
Cat. No.431-2603Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 KD - 18 GT 25 digital, T 25 easy clean digital/control16,6 m/s18,0/12,7 mm194 mm10 - 1500 ml1 * 1 ST1 810,00 €
Specifications
Cat. No.431-0311Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 KD - 18 G - STT 25 digital, T 25 easy clean digital/control17,5 m/s18,0/13,4 mm194 mm10 - 1500 ml1 * 1 ST1 880,00 €
Specifications
Cat. No.IKAA20002973Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 KD - 25 FT 25 digital, T 25 easy clean digital/control23,6 m/s25,0/18,0 mm194 mm100 - 2000 ml1 * 1 ST2 300,00 €
Specifications
Cat. No.IKAA20002975Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 KD - 25 GT 25 digital, T 25 easy clean digital/control22,2 m/s25,0/17,0 mm194 mm50 - 2000 ml1 * 1 ST2 060,00 €
Specifications
Cat. No.431-0312Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 KD - 25 G - STT 25 digital, T 25 easy clean digital/control26,2 m/s25,0/20,0 mm194 mm50 - 2000 ml1 * 1 ST2 240,00 €
Specifications
Cat. No.431-0313Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 KV -18 GT 25 digital15,9 m/s18,0/12,7 mm270 mm10 - 1500 ml1 * 1 ST2 470,00 €
Specifications
Cat. No.431-2455Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 KV - 25 FT 25 digital22,6 m/s25,0/18,0 mm270 mm100 - 2000 ml1 * 1 ST2 730,00 €
Specifications
Cat. No.431-2461Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 KV - 25 GT 25 digital21,4 m/s25,0/17,0 mm270 mm50 - 2000 ml1 * 1 ST2 600,00 €
Specifications
Cat. No.431-2458Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - Dispersing toolS 25 N - 8 GT 25 digital7,7 m/s8,0/6,1 mm108 mm1 - 50 ml1 * 1 ST1 860,00 €
Specifications
Cat. No.431-2451Immersion depth27 - 85 mmUltimate finenessSuspension: 10 - 50 µm; Emulsion: 1 - 10 µmMaterials in contact with mediumPTFE, AISI 316LMax. temperature180 °C - S 25 N - 8 G - STT 25 digital9,6 m/s8,0/6,1 mm108 mm1 - 50 ml1 * 1 ST1 880,00 €
Specifications
Cat. No.431-0253Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 N - 10 GT 25 digital9,4 m/s10,0/7,5 mm105 mm1 - 100 ml1 * 1 ST1 390,00 €
Specifications
Cat. No.431-2452Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 N - 10 G - STT 25 digital9,4 m/s10,0/7,5 mm105 mm1 - 100 ml1 * 1 ST1 460,00 €
Specifications
Cat. No.431-0254Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 N - 18 GT 25 digital15,9 m/s18,0/12,7 mm194 mm10 - 1500 ml1 * 1 ST1 390,00 €
Specifications
Cat. No.431-2453Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 N - 18 G - STT 25 digital15,9 m/s18,0/12,7 mm194 mm10 - 1500 ml1 * 1 ST1 470,00 €
Specifications
Cat. No.431-0222Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 N - 25 FT 25 digital22,6 m/s25,0/18,0 mm194 mm100 - 2000 ml1 * 1 ST1 880,00 €
Specifications
Cat. No.431-2459Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 N - 25 GT 25 digital21,4 m/s25,0/17,0 mm194 mm50 - 2000 ml1 * 1 ST1 780,00 €
Specifications
Cat. No.431-2456Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 25 N - 25 G - STT 25 digital21,4 m/s25,0/17,0 mm194 mm50 - 2000 ml1 * 1 ST1 880,00 €
Specifications
Cat. No.431-0223Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - Dispersing element for S 50 KG-HHS 50 KG - HHT 50 digital-- mm300 mm1 * 1 ST3 620,00 €
Specifications
Cat. No.431-0249Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 50 KG - HH - G 45 FT 50 digital20,9 m/s45,0/40,5 mm290 mm0,25 - 1000 ml1 * 1 ST6 050,00 €
Specifications
Cat. No.431-0257Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 50 KG - HH - G 45 GT 50 digital18,8 m/s45,0/36,0 mm300 mm0,5 - 20 000 ml1 * 1 ST5 490,00 €
Specifications
Cat. No.431-0255Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 50 KG - HH - G 45 MT 50 digital21,2 m/s45,0/40,5 mm300 mm0,5 - 15 000 ml1 * 1 ST5 850,00 €
Specifications
Cat. No.431-0256Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 50 N - G 45 FT 50 basic20,9 m/s45,0/40,0 mm290 mm0,25 - 10 000 ml1 * 1 ST4 190,00 €
Specifications
Cat. No.431-2569Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 50 N - G 45 GT 50 basic18,8 m/s45,0/36,0 mm300 mm0,5 - 20 000 ml1 * 1 ST3 560,00 €
Specifications
Cat. No.431-2560Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 50 N - G 45 G - STT 50 digital19,9 m/s45,0/38,0 mm300 mm50 - 2000 ml1 * 1 ST3 630,00 €
Specifications
Cat. No.431-0258Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 50 N - G 45 MT 50 basic21,2 m/s45,0/40,5 mm290 mm0,5 - 15 000 ml1 * 1 ST3 990,00 €
Specifications
Cat. No.431-2563Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 50 N - W 65 SKT 50 basic28,8 m/s65,0/55,0 mm~400 mm1000 - 10 000 ml1 * 1 ST4 070,00 €
Specifications
Cat. No.431-2581Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature - S 50 N - W 80 SMKT 50 digital80 mm140-350 mm1000 - 50 000 ml1 * 1 ST3 810,00 €
Specifications
Cat. No.431-2582Immersion depthUltimate finenessMaterials in contact with mediumMax. temperature
Accessories
- DescriptionFor unit
- DescriptionFor unit
- DescriptionFor unit
- DescriptionFor unit
- DescriptionFor unit
- IKAA3754000Silentstream flow breakerS 25 N – 18 G; S 25 N – 25 G; S 25 N – 25 F; S 25 NK – 19 G; S 25 KV – 18 G; S 25 KV – 25 G; S 25 KV – 25 F; S 18 N – 19 G; S 25 N – 18 G - ST;S 25 N – 25 G - ST1 * 1 ST - 28,00 €
- DescriptionFor unit
- DescriptionFor unit
- DescriptionFor unit
- DescriptionFor unit
- DescriptionFor unit