Orders LinkContactUsLinkComponent
Homogeniser, T 10 basic ULTRA-TURRAX®
Supplier:   IKA
Homogeniser, T 10 basic ULTRA-TURRAX®
Supplier:  IKA
Certificates
Homogeniser, T 10 basic ULTRA-TURRAX®
1 Options Available Below
Product Details & Documents
Compact, hand-held tissue homogenizer for volumes from 0,5 to 100 ml with a quick-connect coupling for easy interchangeability of dispersing tools.
  • Quick-connect coupling for changing dispersing elements easily
  • Ideal for manual operation due to its light weight and ergonomic design
  • Extremely mobile due to direct line power (no transformer required)
  • Stainless steel dispersing elements can be cleaned quickly and easily as they can be dismounted without tools
  • Plastic disposable dispersing elements are available in two sizes, particularly suitable for PCR analysis
  • High performance drive ensures immense speed stability
  • Quiet operation
Small dispersing instrument for volumes from 0,5 to 100 ml (H₂O). It offers a wide speed range from 8000 to 30,000 min⁻¹ that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements guarantees a wide range of applications.
 : EN 61010-1:2010 + A1:2019 + A1:2019/AC:2019 
EN 61010-2-051:2015 
EN 61326-1:2013 
EN 60529:1991 + A1:2000 + A2:2013 
EN ISO 12100:2010 
2014/35/EU Low Voltage Directive 
2006/42/EG Machinery Directive 
2014/30/EU EMC Directive 
2011/65/EU RoHS Directive
 : The drive unit always requires a dispersing tool, please choose from the compatible tools.
 : The dispersing tool is not included in the scope of delivery. Packages include Stands and Dispersing tools.
 : Supplied with T 10 basic ULTRA-TURRAX® and R 200 clamp.
Specifications for Product Family
Specifications
  • Catalog No:
  • 431-0179
  • 431-0188
  • 431-0189
  • Model:
  • T 10 basic
  • T 10 basic
  • T 10 basic
  • Volume H₂O:
  • 0,5 - 100 ml
  • 0,5 - 100 ml
  • 0,5 - 100 ml
  • Max. viscosity:
  • 5000 mPas
  • 5000 mPas
  • 5000 mPas
  • Speed range:
  • 8000 - 30000 min⁻¹
  • 8000 - 30000 min⁻¹
  • 8000 - 30000 min⁻¹
  • Speed display:
  • Scale
  • Scale
  • Scale
  • Motor input/output:
  • 125/75 W
  • 125/75 W
  • 125/75 W
  • Weight:
  • 0,5 kg
  • 0,5 kg
  • 0,5 kg
  • W×D×H:
  • 56×66×178 mm
  • 56×66×178 mm
  • 56×66×178 mm
Specifications
Product Family Options
T 10 basic ULTRA-TURRAX® Dispersing drive unit
Product Information
  • Availability
    Price
  • 431-0179
    T 10 basic ULTRA-TURRAX® homogeniser, EU-plug
    T 10 basic ULTRA-TURRAX® homogeniser, EU-plug
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 610,00 €
    1 610,00 €
    1 * 1 ST
       1 610,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    431-0179
    Model
    T 10 basic
    Volume H₂O
    0,5 - 100 ml
    Max. viscosity
    5000 mPas
    Speed range
    8000 - 30000 min⁻¹
    Speed display
    Scale
    Motor input/output
    125/75 W
    Weight
    0,5 kg
    W×D×H
    56×66×178 mm
  • 431-0188
    T 10 basic ULTRA-TURRAX® homogeniser, UK-plug
    T 10 basic ULTRA-TURRAX® homogeniser, UK-plug
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    431-0188
    Model
    T 10 basic
    Volume H₂O
    0,5 - 100 ml
    Max. viscosity
    5000 mPas
    Speed range
    8000 - 30000 min⁻¹
    Speed display
    Scale
    Motor input/output
    125/75 W
    Weight
    0,5 kg
    W×D×H
    56×66×178 mm
  • 431-0189
    T 10 basic ULTRA-TURRAX® homogeniser, CH-plug
    T 10 basic ULTRA-TURRAX® homogeniser, CH-plug
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    431-0189
    Model
    T 10 basic
    Volume H₂O
    0,5 - 100 ml
    Max. viscosity
    5000 mPas
    Speed range
    8000 - 30000 min⁻¹
    Speed display
    Scale
    Motor input/output
    125/75 W
    Weight
    0,5 kg
    W×D×H
    56×66×178 mm
Dispersing tools
Product Information
  • Catalog #
    Volume
    Stator/rotor Ø
    Speed
    For
    Length
    Description
    Availability
    Price
  • 1 - 40 ml
    7 / 4,8 mm
    7,5 m/s
    T 10 basic
    110 mm
    S10D-7G-KS-110
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 627,00 €
    627,00 €
    1 * 24 ST
       627,00 €
Product Information
  • Catalog #
    Length
    Stator/rotor Ø
    Speed
    For
    Volume
    Description
    Availability
    Price
  • 115 mm
    10/7,6 mm
    11,9 m/s
    T 10 basic
    1 - 100 ml
    S 10 N - 10 G
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 560,00 €
    1 560,00 €
    1 * 1 ST
       1 560,00 €
Product Information
  • Catalog #
    Length
    Stator/rotor Ø
    Speed
    For
    Volume
    Description
    Availability
    Price
  • 115 mm
    10,0/7,6 mm
    11,9 m/s
    T 10 basic
    1 - 100 ml
    S 10 N - 10 G - ST
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 690,00 €
    1 690,00 €
    1 * 1 ST
       1 690,00 €
Product Information
  • Catalog #
    Volume
    Stator/rotor Ø
    Speed
    For
    Length
    Description
    Availability
    Price
  • 1 - 20 ml
    7 / 4,8 mm
    7,5 m/s
    T 10 basic
    65 mm
    S10D-7G-KS-65
    Log in for availability
    Log in for availability
    1 * 24 ST
    1 * 24 ST - 555,00 €
    555,00 €
    1 * 24 ST
       555,00 €
Product Information
  • Catalog #
    Length
    Stator/rotor Ø
    Speed
    For
    Volume
    Description
    Availability
    Price
  • 115 mm
    8/6,1 mm
    9,6 m/s
    T 10 basic
    1 - 50 ml
    S 10 N - 8 G
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 560,00 €
    1 560,00 €
    1 * 1 ST
       1 560,00 €
Product Information
  • Catalog #
    Length
    Stator/rotor Ø
    Speed
    For
    Volume
    Description
    Availability
    Price
  • 115 mm
    8,0/6,1 mm
    9,6 m/s
    T 10 basic
    1 - 50 ml
    S 10 N - 8 G - ST
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 690,00 €
    1 690,00 €
    1 * 1 ST
       1 690,00 €
Product Information
  • Catalog #
    Length
    Stator/rotor Ø
    Speed
    For
    Volume
    Description
    Availability
    Price
  • 92 mm
    5/3,8 mm
    6 m/s
    T 10 basic
    0,5 - 10 ml
    S 10 N - 5 G
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 560,00 €
    1 560,00 €
    1 * 1 ST
       1 560,00 €
Stands
Product Information
  • Catalog #
    For
    Description
    Availability
    Price
  • T 10 basic/VDI 12
    Plate stand, 242×355 mm, rod 10 mm Ø with height 370 mm. Max. load: 0,7 kg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 208,00 €
    208,00 €
    1 * 1 ST
       208,00 €
Product Information
  • Catalog #
    For
    Description
    Availability
    Price
  • 3
    Plate stand R 1825, 200×316 mm, rod 16 mm Ø with height 560 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 355,00 €
    355,00 €
    1 * 1 ST
       355,00 €
Product Information
  • Catalog #
    For
    Description
    Availability
    Price
  • Plate stand, 200×316 mm, rod 16 mm Ø with height 800 mm
    Plate stand R 1826, plate 200x316 mm, rod 800 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 392,00 €
    392,00 €
    1 * 1 ST
       392,00 €
Product Information
  • Catalog #
    For
    Description
    Availability
    Price
  • Plate stand, 200×316 mm, rod 16 mm Ø with height 1000 mm
    Plate stand R 1827
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 418,00 €
    418,00 €
    1 * 1 ST
       418,00 €
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • R200 clamp for fastening the T 10 basic to the stand
    R200 clamp for fastening the T 10 basic to the stand
    442-0313
    T 10 basic
    Log in for availability
    1 * 1 ST - 45,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Bosshead clamp
    Bosshead clamp
    442-2043
    Log in for availability
    1 * 1 ST - 42,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Beaker cap, BC 1000, silicone, for 600 and 1000 ml beakers
    Beaker cap, BC 1000, silicone, for 600 and 1000 ml beakers
    IKAA20003417
    Log in for availability
    1 * 1 ST - 22,50 €