ContactUsLinkComponent
Furniture fixed under bench and base trim
Furniture fixed under bench and base trim
Catalog # 132-0279
Supplier:  EQUIP LABO
undefined
Furniture fixed under bench and base trim
Catalog # 132-0279
Supplier:  EQUIP LABO
Specifications
  • Pk:
    1
Specifications
About this item
Furniture fixed (delivered without bench).
The length of the furniture corresponds to the length of the bench.
The depth of the furniture corresponds to the depth of the bench counter-top (excluding splash back and shelf). In the case of central benches or benches not attached to a wall, a base trim provides a better finish.
Colours: grey facade, white handles, white sides, white base.
Delivery: Delivered with a removable shelf as standard (possibility of additional shelves on request).
Caution: It is essential that this furniture is mounted with a bench.