Distillation and separation apparatus, behrotest® Kompact
Supplier: BEHR
Certificates
About this item
Complete sets of equipment for preparation and distillation for commonly required analyses of solid and aqueous samples.
- Simple and convenient to use: All apparatus is delivered already mounted on stands with tubing connections pre-installed - no more tedious fiddling with clamps and mountings
- Space saving: All apparatus is mounted on a compact 23×27 cm base plate, including the units for dual sampling
- Analytical reliability: All components comply with the applicable official analytical procedures and safety standards
Frequently Bought Together