Order Entry
Belgium
ContactUsLinkComponent
behrotest® DET supplementary set for Soxhlet extraction (30 ml), DE 30
behrotest® DET supplementary set for Soxhlet extraction (30 ml), DE 30
  705-0026
 :  BEHR
undefined
behrotest® DET supplementary set for Soxhlet extraction (30 ml), DE 30
  705-0026
 :  BEHR

 

  • Type:
    DE 30
  • Description:
    Supplementary set for Soxhlet Extraction (30 ml)
  • Pk:
    1

 

 

This set includes the following equipment:
5×Sample vessels, 250 ml 65/250
5×Vessel –extractor connector M 2965
5×30 ml soxhlet extractor EZ 30
5×Reflux condenser for EZ 30 extractor RFK 30

 : The DET Basic unit (DBAS, 705-0023) is also required.