ContactUsLinkComponent
Accessories for electroporation system, ECM® 830
Accessories for electroporation system, ECM® 830
Catalog # 732-0026
Supplier:  BTX
undefined
Accessories for electroporation system, ECM® 830
Catalog # 732-0026
Supplier:  BTX
Specifications
  • Description:
    Footswitch for legacy ECM® 830, model 1250 FS
  • Pk:
    1
Specifications
About this item
Footswitch for legacy ECM® 830 (serial numbers of the compatible units: 8310249 to date).