Well visualisation plates
Supplier: EMBITEC
Jump to:
About this item
RunOne™ well visualisation plates are designed to make gel loading simple. The light colour of most gel units provides little or no contrast for the agarose gel and the gel wells. RunOne™ well visualisation plates are red, fit securely into the RunOne™ system, and provide a stable platform for gel running.
2 Options Available Below
Product Details & Documents
RunOne™ well visualisation plates are designed to make gel loading simple. The light colour of most gel units provides little or no contrast for the agarose gel and the gel wells. RunOne™ well visualisation plates are red, fit securely into the RunOne™ system, and provide a stable platform for gel running.
Product Family Options
Product Information
- AvailabilityPrice
- 730-2203Well visualisation plate, 9.5×11.6 cm, with 2 rails for locking medium gels in placeWell visualisation plate, 9.5×11.6 cm, with 2 rails for locking medium gels in placeLog in for availability1 * 1 ST14,60 €
- 730-2202Well visualisation plate, 9.5×11.6 cm, with 4 rails for locking portrait and landscape gels in placeWell visualisation plate, 9.5×11.6 cm, with 4 rails for locking portrait and landscape gels in placeLog in for availability1 * 1 ST14,60 €