Gel casting system, MultiCaster
: EMBITEC
Moulded PC. Compatible with the RunOne™ electrophoresis system.
- Simple to use - no gaskets, clamps or tape required
- Flexible with options for well size, well configuration, number of wells and resolving area
Moulded PC. Compatible with the RunOne™ electrophoresis system.
- Simple to use - no gaskets, clamps or tape required
- Flexible with options for well size, well configuration, number of wells and resolving area
- Catalog No:
- 700-4209
- 700-4211
- 700-4212
- 700-4210
- 730-0360
- Gel dimensions:
- 115×119
- 115×89
- 115×119
- 115×89
- —
Product Family Options
- Description
- MultiCaster, blue casting stand, UVT tray, 6 reversible combs and a red plate, 8-channel pipette compatible1 * 1 ST421,00 €
Specifications
Cat. No.700-4209Gel dimensions115×119 - MultiCaster, orange casting stand, UVT tray, 2 reversible combs and a red plate1 * 1 ST278,00 €
Specifications
Cat. No.700-4211Gel dimensions115×89 - MultiCaster, turquoise casting stand, UVT tray, 4 reversible combs and a red plate, 12-channel pipette compatible1 * 1 ST421,00 €
Specifications
Cat. No.700-4212Gel dimensions115×119 - MultiCaster, yellow casting stand, UVT tray, 8 reversible combs, and a red plate, 12-channel pipette compatible1 * 1 ST421,00 €
Specifications
Cat. No.700-4210Gel dimensions115×89 Specifications
Cat. No.730-0360Gel dimensions—
Combs
- Description