ContactUsLinkComponent
Multiple horizontal gel electrophoresis system, Owl™ A2-OK
Multiple horizontal gel electrophoresis system, Owl™ A2-OK
Catalog # 730-0161
undefined
Multiple horizontal gel electrophoresis system, Owl™ A2-OK
Catalog # 730-0161
Specifications
  • Model:
    Owl A2-OK
  • Description:
    Multiple gel system
  • Buffer volume:
    2300 ml
  • Gel size W×L:
    70×80 mm
  • Unit W×L×H:
    280×375×100 mm
  • Pk:
    1
Specifications
About this item
Owl™ A2-OK multiple gel system is ideal for teaching labs or labs that need to run multiple gels simultaneously.
  • Up to six mini gels can be run
  • One teacher can instruct up to 12 students with one system and only one power supply
  • The system can screen up to 144 samples at once when used with optional combs (120 with supplied combs)
  • EasyCast casting design allows the casting of gels without tape or plugs
  • UV transmissible gel trays may be placed directly on the transilluminator for photodocumentaion
Delivery: Each unit includes buffer chamber with 3-point levelling base, SuperSafe lid with attached power supply leads, EasyCast gasketed UVT gel trays (6), external casting chambers (2), gel tray holder, and 12 double-sided combs (10-well, 1,0/1,5 mm thick).
Combs
Product Information