ContactUsLinkComponent
pH/mV/°C/ion meter, bench, inoLab® pH/ION 7320
 :  WTW
undefined
pH/mV/°C/ion meter, bench, inoLab® pH/ION 7320
 :  WTW
  • Catalog No:
  • 662-1685
  • 662-1683
  • 662-1684
  • 662-1686
  • Model:
  • pH/ION 7320
  • pH/ION 7320
  • pH/ION 7320
  • pH/ION 7320
  • pH range:
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • pH resolution:
  • 0,01 / 0,001
  • 0,01 / 0,001
  • 0,01 / 0,001
  • 0,01 / 0,001
  • pH accuracy:
  • ±0,005 / ±0,01
  • ±0,005 / ±0,01
  • ±0,005 / ±0,01
  • ±0,005 / ±0,01
  • pH calibration:
  • MultiCal® automatic calibration (1 - 5 points)
  • MultiCal® automatic calibration (1 - 5 points)
  • MultiCal® automatic calibration (1 - 5 points)
  • MultiCal® automatic calibration (1 - 5 points)
  • mV range:
  • ±1200,0 / ±2500
  • ±1200,0 / ±2500
  • ±1200,0 / ±2500
  • ±1200,0 / ±2500
  • mV accuracy:
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ±0,3 / ±1
  • ISE range:
  • 0 - 999999
  • 0 - 999999
  • 0 - 999999
  • 0 - 999999
  • ISE resolution:
  • 1, 2 or 3 significant digits
  • 1, 2 or 3 significant digits
  • 1, 2 or 3 significant digits
  • 1, 2 or 3 significant digits
  • ISE calibration:
  • Automatic calibration (1 - 7 points)
  • Automatic calibration (1 - 7 points)
  • Automatic calibration (1 - 7 points)
  • Automatic calibration (1 - 7 points)
  • ISE units:
  • mg/l, mg/kg, mmol/l, ppm, %
  • mg/l, mg/kg, mmol/l, ppm, %
  • mg/l, mg/kg, mmol/l, ppm, %
  • mg/l, mg/kg, mmol/l, ppm, %
  • Temperature range:
  • −5...+105 °C
  • −5...+105 °C
  • −5...+105 °C
  • −5...+105 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • ±0,1 °C
  • Display:
  • Backlit graphic display with CMC
  • Backlit graphic display with CMC
  • Backlit graphic display with CMC
  • Backlit graphic display with CMC
  • Compliance:
  • IP 43, GLP
  • IP 43, GLP
  • IP 43, GLP
  • IP 43, GLP
  • Data logging capacity:
  • Manual (500), automatic (5000), programmable
  • Manual (500), automatic (5000), programmable
  • Manual (500), automatic (5000), programmable
  • Manual (500), automatic (5000), programmable
  • Connections:
  • DIN or BNC
  • DIN or BNC
  • DIN or BNC
  • DIN or BNC
  • Outputs:
  • USB
  • USB
  • USB
  • USB
Frequently Bought Together