Orders LinkContactUsLinkComponent
Conductivity cells
Conductivity cells
Supplier:   WTW
Conductivity cells
Supplier:  WTW
Certificates
Conductivity cells
Conductivity cells
11 Options Available Below
Product Details & Documents
TetraCon® 325, TetraCon® V and KLE 325 are suitable for on-site measurements in rivers, lakes and wastewater, fish farming, ground water measurements and applications in water laboratories.
TetraCon® 325/C is ideal for heavy chemical applications, e.g. for acid media.
TetraCon® 325/S is great for measurements in pastes, creams and emulsions.
LR 325/01 is good for measurements in ultapure water.
Product Family Options
Product Information
  • Type
    Conductivity
    Cell constant
    Temp.
    Material
    Ø×L
    Connection
    Availability
    Price
  • 663-8118
    LR 325/01 with integrated temp. sensor, for ultrapure water
    LR 325/01 with integrated temp. sensor, for ultrapure water
    LR 325/01
    0,001 - 200 µS/cm
    0,100 cm-1
    –5...+80 °C
    Stainless steel
    12×120 mm
    1,5 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 838,00 €
    838,00 €
    1 * 1 ST
       838,00 €
  • 662-0484
    LR 325/001 with integrated temp. sensor
    LR 325/001 with integrated temp. sensor
    LR 325/001
    0,0001 - 30 µS/cm
    0,01 cm-1
    0...100 °C
    Stainless steel
    20×120 mm
    1,5 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 400,00 €
    1 400,00 €
    1 * 1 ST
       1 400,00 €
  • 662-0481
    TetraCon® 325/S, POM head, immersion depth 40/120 mm
    TetraCon® 325/S, POM head, immersion depth 40/120 mm
    TetraCon® 325/S, immersion depth 40/120 mm
    1 µS/cm - 2 S/cm
    0,491 cm-1
    0...100 °C
    Epoxy
    15,3×120 mm
    1,5 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 920,00 €
    920,00 €
    1 * 1 ST
       920,00 €
  • 663-8117
    TetraCon® 325, POM head, immersion depth 36/120 mm, with integrated temp. sensor
    TetraCon® 325, POM head, immersion depth 36/120 mm, with integrated temp. sensor
    TetraCon® 325, POM head, immersion depth 36/120 mm
    1 µS/cm - 2 S/cm
    0,475 cm-1
    0...100 °C
    Epoxy
    15,3×120 mm
    1,5 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 675,00 €
    675,00 €
    1 * 1 ST
       675,00 €
  • 663-0227
    TetraCon® 325/C, for chemical media
    TetraCon® 325/C, for chemical media
    TetraCon® 325/C
    1 µS/cm - 2 S/cm
    0,475 cm-1
    –5...+80 °C
    Epoxy
    15,3×120 mm
    1,5 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 010,00 €
    1 010,00 €
    1 * 1 ST
       1 010,00 €
  • 663-1841
    TetraCon® 325-3, POM head, with integrated temp. sensor
    TetraCon® 325-3, POM head, with integrated temp. sensor
    TetraCon® 325-3, POM head
    1 µS/cm - 2 S/cm
    0,475 cm-1
    –5...+80 °C
    Epoxy
    15,3×120 mm
    3 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 720,00 €
    720,00 €
    1 * 1 ST
       720,00 €
  • 663-1842
    TetraCon® 325-6, POM head, with integrated temp. sensor
    TetraCon® 325-6, POM head, with integrated temp. sensor
    TetraCon® 325-6, POM head
    1 µS/cm - 2 S/cm
    0,475 cm-1
    –5...+80 °C
    Epoxy
    15,3×120 mm
    6 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 897,00 €
    897,00 €
    1 * 1 ST
       897,00 €
  • 663-1843
    TetraCon® 325-10, POM head, with integrated temp. sensor
    TetraCon® 325-10, POM head, with integrated temp. sensor
    TetraCon® 325-10, POM head
    1 µS/cm - 2 S/cm
    0,475 cm-1
    –5...+80 °C
    Epoxy
    15,3×120 mm
    10 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 972,00 €
    972,00 €
    1 * 1 ST
       972,00 €
  • 663-1844
    TetraCon® 325-15, POM head, with integrated temp. sensor
    TetraCon® 325-15, POM head, with integrated temp. sensor
    TetraCon® 325-15, POM head
    1 µS/cm - 2 S/cm
    0,475 cm-1
    –5...+80 °C
    Epoxy
    15,3×120 mm
    15 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 030,00 €
    1 030,00 €
    1 * 1 ST
       1 030,00 €
  • 663-1845
    TetraCon® 325-20, POM head, with integrated temp. sensor
    TetraCon® 325-20, POM head, with integrated temp. sensor
    TetraCon® 325-20, POM head
    1 µS/cm - 2 S/cm
    0,475 cm-1
    –5...+80 °C
    Epoxy
    15,3×120 mm
    20 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 160,00 €
    1 160,00 €
    1 * 1 ST
       1 160,00 €
  • 663-0146
    KLE 325 with integrated temp. sensor
    KLE 325 with integrated temp. sensor
    KLE 325
    10 µS/cm - 20 mS/cm
    0,84 cm-1
    0...80 °C
    Epoxy
    15,3×120 mm
    1,5 m cable
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 563,00 €
    563,00 €
    1 * 1 ST
       563,00 €
Flow through vessel
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Flow-through vessel, glass, D01/T, for LR 325/01
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 266,00 €
    266,00 €
    1 * 1 ST
       266,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Flow vessel for multi-parameter measurements, D 700/N, can be populated with TriOxmatic® 700 IQ, SensoLyt® SE/DW/EC/platinum, TetraCon® 325 and TFK 325
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 480,00 €
    1 480,00 €
    1 * 1 ST
       1 480,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Replacement flow through vessel, D 001, for LR 325/001
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 417,00 €
    417,00 €
    1 * 1 ST
       417,00 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Flow-through vessel, transparent PVC, i-Ø 18 mm, D 1/T, for old measuring cells
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 690,00 €
    690,00 €
    1 * 1 ST
       690,00 €