Orders LinkContactUsLinkComponent
Pascal pressure device
Pascal pressure device
Supplier:   BREUKHOVEN
Pascal pressure device
Supplier:  BREUKHOVEN
Certificates
Pascal pressure device
Pascal pressure device
2 Options Available Below
Product Details & Documents
Steel stand. Scale may be read from a distance. Rubber membrane.
  • Counterweight needle
  • Level needle
  • 4 jugs with cone
Product Family Options
Product Information
  • Availability
    Price
  • 761-0059
    apparatus
    apparatus
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 761-0048
    Replacement membrane in rubber
    Replacement membrane in rubber
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST