Multiplace Kjeldahl digestion units
Supplier: GERHARDT
Certificates
About this item
Traditional Kjeldahl digestion units for flask sizes from 50 to 750 ml.
- Adjustable temperature
- Up to 600 °C for the type KI8/16, power 1800 W
- Up to 650 °C for the type KI11/26 400 V, 3 phase, power 3480 W
Delivery information: Supplied with Kjeldahl flasks, glass exhaust manifold, forked support rods, and power cord but without water jet pump.
Frequently Bought Together