ContactUsLinkComponent
Software, Liqui-Soft® 2.0
Software, Liqui-Soft® 2.0
Catalog # 612-9837
Supplier:  HIRSCHMANN
Software, Liqui-Soft® 2.0
Catalog # 612-9837
Supplier:  HIRSCHMANN
Specifications
  • Description:
    Software, liqui-soft® 2.0
  • Pk:
    1
Specifications
About this item
Software for titration, distribution, documentation, and automatic calibration.
  • Enables up to four akku-drive®, solarus® or other types of dispensers to be controlled by the PC
  • Very user-friendly interface, simple protocol documentation
  • Any volumetric measurement instrument can be calibrated and the data required for compliance with DIN and ISO standards are stored in memory
  • Simultaneous recording and storage of all measured values
  • Compatible with Windows 95, 98, ME, NT, 2000 and XP