ContactUsLinkComponent
Distillation, extraction and thermal reactions, behrotest® DET systems
Distillation, extraction and thermal reactions, behrotest® DET systems
  705-0023
 :  BEHR
undefined
Distillation, extraction and thermal reactions, behrotest® DET systems
  705-0023
 :  BEHR
  • Description:
    DET basic unit
  • Pk:
    1
  • Type:
    DBAS
The behrotest® universal analytical DET system is a flexible modular system designed for laboratories with frequently changing applications who do not want to invest in dedicated complete apparatus for each application. The system has a core or basic unit to which application specific supplementary/expansion sets can be added. The expansion kits comprise typically of the additional specific glassware and gas flow connections.
The behrotest® DET DBAS includes the following equipment: