Orders LinkContactUsLinkComponent
Manual cell counters
Manual cell counters
Supplier:   MOINEAU
Manual cell counters
Supplier:  MOINEAU
Certificates
Manual cell counters
Manual cell counters
5 Options Available Below
Product Details & Documents
This manual cell counter can be used in combination of up to five counters.
  • Four digit display (4,3×3,5 mm) with reset knob
  • Use finger pressure
  • Can be fixed in position
W×D×H: 37,6×0,4×38,1 mm
Product Family Options
Product Information
  • Array
    Availability
    Price
  • MOIN1494MW
    Manual cell counter
    Manual cell counter
    4
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 402,00 €
    402,00 €
    1 * 1 ST
       402,00 €
  • MOIN1495/2MW
    Manual cell counter
    Manual cell counter
    10
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 793,00 €
    793,00 €
    1 * 1 ST
       793,00 €
  • 720-0162
    Manual cell counter
    Manual cell counter
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,70 €
    51,70 €
    1 * 1 ST
       51,70 €
  • 720-0163
    Manual cell counter
    Manual cell counter
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 215,00 €
    215,00 €
    1 * 1 ST
       215,00 €
  • 720-0164
    Manual cell counter
    Manual cell counter
    5
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 494,00 €
    494,00 €
    1 * 1 ST
       494,00 €