Orders LinkContactUsLinkComponent
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
Supplier:   Lovibond Tintometer
Comparator kits, Lovibond® system 2000
Certificates
Comparator kits, Lovibond® system 2000
Comparator kits, Lovibond® system 2000
8 Options Available Below
Product Details & Documents
Available for many of the commonly used colour scales, these kits are a convenient means of ordering the complete range of equipment required. Each system includes.
  • A visual comparator instrument which brings the sample and colour standards into the same field of view for more accurate colour matching
  • A selection of colour standards - stable-coloured glass filters in test discs, which have been pre-calibrated to give direct readings over set measuring ranges
  • A standardised benchtop light source to ensure constant, uniform lighting conditions for colour matching and, therefore, consistent and accurate readings
  • Glassware of the appropriate type and path length
Product Family Options
Product Information
  • Type
    Availability
    Price
  • 633-0022
    Platinum-Cobalt Units (Low Range, 0 - 70 mg Pt/l)) Nessleriser 2250 with Daylight 2000 Lighting unit and 250 mm Nessler cylinders; Pt-Co discs CAA (0 - 30 mg Pt/l), CAB (30 - 70 mg Pt/l); stand for using Nessleriser with natural lighting
    Platinum-Cobalt Units (Low Range, 0 - 70 mg Pt/l)) Nessleriser 2250 with Daylight 2000 Lighting unit and 250 mm Nessler cylinders; Pt-Co discs CAA (0 - 30 mg Pt/l), CAB (30 - 70 mg Pt/l); stand for using Nessleriser with natural lighting
    AF328
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 270,00 €
    2 270,00 €
    1 * 1 ST
       2 270,00 €
  • 633-0024
    Gardner Colour Comparator 2000+ with Daylight 2000 Lighting unit; Gardner discs: 4/30 AS, 1 - 9; 4/30 BS, 10 - 18; W680/OG/10 mm fused cell, with 10 mm path length
    Gardner Colour Comparator 2000+ with Daylight 2000 Lighting unit; Gardner discs: 4/30 AS, 1 - 9; 4/30 BS, 10 - 18; W680/OG/10 mm fused cell, with 10 mm path length
    AF334
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 290,00 €
    2 290,00 €
    1 * 1 ST
       2 290,00 €
  • 633-0023
    European Pharmacopoeia (EP) Colour Comparator 2000+ with Daylight 2000 Lighting unit; EP discs: 4/64A, R1-R7; 4/64B, Y1-Y7; 4/64C, B1-B9; 4/64D, BY1-BY7; 4/64D, GY1-GY7; 2 x W680/OG/40 mm fused cells, with 40 mm path length
    European Pharmacopoeia (EP) Colour Comparator 2000+ with Daylight 2000 Lighting unit; EP discs: 4/64A, R1-R7; 4/64B, Y1-Y7; 4/64C, B1-B9; 4/64D, BY1-BY7; 4/64D, GY1-GY7; 2 x W680/OG/40 mm fused cells, with 40 mm path length
    AF332
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 550,00 €
    3 550,00 €
    1 * 1 ST
       3 550,00 €
  • 633-0152
    IFU colour scale colour grading kit for fruit juices, including Colour Comparator 2000+ with Daylight 2000 Lighting unit,FU discs 4/48,1 - 9; 4/49 9 - 17; 4/50 17 - 25), W680/OG/13 mm fused cells
    IFU colour scale colour grading kit for fruit juices, including Colour Comparator 2000+ with Daylight 2000 Lighting unit,FU discs 4/48,1 - 9; 4/49 9 - 17; 4/50 17 - 25), W680/OG/13 mm fused cells
    AF331
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 690,00 €
    2 690,00 €
    1 * 1 ST
       2 690,00 €
  • 633-0153
    Iodine colour scale with Comparator 2000+ with Daylight 2000 Lighting unit, iodine discs, W680/OG/25 and W680/OG/10 mm fused cells
    Iodine colour scale with Comparator 2000+ with Daylight 2000 Lighting unit, iodine discs, W680/OG/25 and W680/OG/10 mm fused cells
    AF333
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 820,00 €
    2 820,00 €
    1 * 1 ST
       2 820,00 €
  • 633-0021
    Platinum-Cobalt Units (10 - 250 mg Pt/l) Nessleriser 2150 with Daylight 2000 Lighting unit and 113 mm (50 ml) Nessler cylinders; Pt-Co discs NSH (10 - 90 mg Pt/l), NSB (70 - 250 mg Pt/l); stand for using Nessleriser with natural lighting
    Platinum-Cobalt Units (10 - 250 mg Pt/l) Nessleriser 2150 with Daylight 2000 Lighting unit and 113 mm (50 ml) Nessler cylinders; Pt-Co discs NSH (10 - 90 mg Pt/l), NSB (70 - 250 mg Pt/l); stand for using Nessleriser with natural lighting
    AF325
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 230,00 €
    2 230,00 €
    1 * 1 ST
       2 230,00 €
  • 633-0151
    European Beer Convention (EBC) colour scale for measuring beers and malts, Comparator 2000+ with Daylight 2000 Lighting unit (EBC version), EBC discs, W680/OG/10 mm fused cells
    European Beer Convention (EBC) colour scale for measuring beers and malts, Comparator 2000+ with Daylight 2000 Lighting unit (EBC version), EBC discs, W680/OG/10 mm fused cells
    AF330
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 750,00 €
    2 750,00 €
    1 * 1 ST
       2 750,00 €
  • 633-0259
    USP colour scale with Comparator 2000+ with Daylight 2000 Lighting unit, USP discs: 4/66 A, 4/66 B, 4/66 C, 4/66 D; 2 × W680/OG/40 mm fused cell, with 40 mm path length
    USP colour scale with Comparator 2000+ with Daylight 2000 Lighting unit, USP discs: 4/66 A, 4/66 B, 4/66 C, 4/66 D; 2 × W680/OG/40 mm fused cell, with 40 mm path length
    AF335
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 390,00 €
    2 390,00 €
    1 * 1 ST
       2 390,00 €