Orders LinkContactUsLinkComponent
Labwise® software
Labwise® software
Labwise® Control and Analysis software. Includes USB & RS232 cable, GR150, GP200, TX150, TXF200
Catalog # 461-0223
Supplier:   GRANT INSTRUMENTS
Labwise® software
Catalog # 461-0223
Supplier:  GRANT INSTRUMENTS
Labwise® software
Labwise® software
Labwise® Control and Analysis software. Includes USB & RS232 cable, GR150, GP200, TX150, TXF200
Price
175,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The high performance GR150, GP200 and new TX150 and TXF200 thermostatic controllers together with Labwise® software make set up, monitoring and data logging straightforward. Labwise® features a graphic display of the controller's performance as a program is run, has real time zoom and scaling of graphical display and allows data to be logged for future recall and analysis. Labwise® software controls the pump speed for GP200/TXF200 series units and runs on a PC. User has choice of set up language: English, French, German, Spanish or Italian.
Set up features include:
Set temperature, high and low alarms; alarms can be configured to switch a relay
Set reaction timer and delayed start and stop time
Control of output relays for operating ancillary equipment.
Programming features include:
Program values may be set graphically or numerically with up to 30 segments per program
Set heat or cool time to target and set number of loops, 1 to 254, or infinite looping between selected points.
Specifications
  • Description:
    Labwise® Control and Analysis software. Includes USB & RS232 cable
  • Seq:
    15
  • For:
    GR150, GP200, TX150, TXF200