ContactUsLinkComponent
Certificate of calibration
Certificate of calibration
Catalog # 620-1494
Supplier:  DICKSON
Certificate of calibration
Catalog # 620-1494
Supplier:  DICKSON
Specifications
  • Description:
    One-point NIST Traceable® calibration certificate
  • Pk:
    1
  • For:
    Chart recorders
Specifications
About this item
A NIST certificate provides that your Dickson instrument had been properly calibrated and is traceable back to the National Institute of Standards and Technology (NIST).
  • Includes documentation to one Dickson pre-selected point after calibration
  • Allows you to comply with audits
  • Helps keep track of instrument calibration requirements
  • Easy recalibration ordering information also included
This type of documented calibration will satisfy most quality and compliance situations. This one-point NIST offers proof of the accuracy of Dickson''s quality instruments.