Orders LinkContactUsLinkComponent
Mini Coolers
Mini Coolers
Supplier:   Brand
Mini Coolers
Supplier:  Brand
Certificates
Mini Coolers
3 Options Available Below
Product Details & Documents
PC, yellow, with clear lid. Maintains temperature of 0 °C for 60 min.
  • Durable polycarbonate filled with non-toxic gel
  • Mini coolers hold twelve 0,5 to 2,0 ml tubes
Mini coolers are designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.
Product Family Options
Product Information
  • For
    No. of Holes
    Colour
    Availability
    Price
  • 479-1241
    Mini Cooler, maintains the Temperature at 0 °C during 60 Minutes
    Mini Cooler, maintains the Temperature at 0 °C during 60 Minutes
    0,5 to 2,0 ml tubes
    12
    Red
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 126,00 €
    126,00 €
    1 * 1 ST
       126,00 €
  • 479-1242
    Mini cooler, maintains the Temperature at −20 °C during 60 Minutes
    Mini cooler, maintains the Temperature at −20 °C during 60 Minutes
    0,5 to 2,0 ml tubes
    12
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 126,00 €
    126,00 €
    1 * 1 ST
       126,00 €
  • 479-1243
    Mini cooler, maintains the Temperature at −70 °C during 45 Minutes
    Mini cooler, maintains the Temperature at −70 °C during 45 Minutes
    0,5 to 2,0 ml tubes
    12
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 126,00 €
    126,00 €
    1 * 1 ST
       126,00 €