VWR® AVS TITRINORM® Potassium Chloride Electrolytic Solutions
Supplier: VWR Chemicals
Certificates
About this item
Potassium chloride 3 Molar solution may also be used as an electrode storage solution.
For full information concerning health and safety data please see the SDS on the website.
Related Information
Certificates
Search by Lot Number100 ml plastic bottle for liquids DIN 28 (ZPACK13948098) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 500 ml Plastic bottle for liquids DIN 45 (ZPACK13960500)
Frequently Bought Together