pH electrodes for special laboratory applications
Supplier: HAMILTON BONADUZ
Jump to:
About this item
Electrodes for special laboratory applications such as microbiological samples, serum, flat surfaces, small volumes, and samples containing fats and proteins.
- Long-term stable Everef system
- Different tip designs for different applications
- Polisolve electrolyte: maintenance-free
- 3M KCl and Protelyte electrolytes: Refillable
pH range: 0 to 14
: Supplied with individual certificate and serial number. Electrodes with S7 connector head are supplied without cable which needs to be ordered separately.
7 Options Available Below
Product Details & Documents
Electrodes for special laboratory applications such as microbiological samples, serum, flat surfaces, small volumes, and samples containing fats and proteins.
- Long-term stable Everef system
- Different tip designs for different applications
- Polisolve electrolyte: maintenance-free
- 3M KCl and Protelyte electrolytes: Refillable
pH range: 0 to 14
: Supplied with individual certificate and serial number. Electrodes with S7 connector head are supplied without cable which needs to be ordered separately.
Product Family Options
Product Information
- TypeTemp.MaterialElectrolyteDiaphr.Ø×LConnectionForAvailabilityPrice
- MiniTrode0...+100 °CGlass3 mol/l KClCeramic3×60 mmS7Small volumes (e.g. vials)Log in for availability1 * 1 ST312,00 €
- BioTrode0...+100 °CGlassProtelyteCeramic3×60 mmS7Small volumes containing proteins (e.g. microtiter plates)Log in for availability1 * 1 ST319,00 €
- SpinTrode0...+100 °CGlass3 mol/l KClCeramic3×180 mmS7Small volumes (e.g. NMR tubes)Log in for availability1 * 1 ST401,00 €
- SlimTrode0...+100 °CGlass3 mol/l KClCeramic6×100 mmS7Measurements in test tubesLog in for availability1 * 1 ST257,00 €
- FlushTrode−10...+80 °CGlass3 mol/l KClSleeve12×120 mmS7Viscous, ion-weak, and proteinaceous mediaLog in for availability1 * 1 ST260,00 €
- FlaTrode0...+60 °CPlasticPolisolveSingle Pore ring12×120 mmS7Surfaces (paper, agar plates, skin)Log in for availability1 * 1 ST262,00 €
- FillTrode0...+60 °CPlasticPolisolveSingle Pore ring12×120 mmS7Multi-use (e.g. viscous samples)Log in for availability1 * 1 ST241,00 €
Accessories
Product Information
- Catalog #PkAvailabilityPrice
- Electrolyte KCl 3 M, 100 ml
Product Information
- Catalog #PkAvailabilityPrice
- Electrolyte Protelyte, 100 ml
Product Information
- Catalog #PkAvailabilityPrice
- Electrode storage solution, 500 ml, for pH and ORP sensorsElectrode storage solution, 500 ml, for pH and ORP sensors662-9223500 mlLog in for availability1 * 500 mL - 32,30 €
Product Information
- Catalog #AvailabilityPrice
- Sensor cable S7/no connector (open end), length 1 mSensor cable S7/no connector (open end), length 1 m662-9229Log in for availability1 * 1 m - 48,50 €
Product Information
- Catalog #AvailabilityPrice
- Sensor cable, S7/BNC, length 3 m
Product Information
- Catalog #AvailabilityPrice
- Sensor cable, S7/DIN, length 3 m
Product Information
- Catalog #AvailabilityPrice
- Sensor cable S7/BNC, length 1 m
Product Information
- Catalog #PkAvailabilityPrice
- pH cleaning solution set (2×500 ml solution A+B + 1×500 ml storage solution)pH cleaning solution set (2×500 ml solution A+B + 1×500 ml storage solution)HAMI2382901 SETLog in for availability1 * 1 SET - 163,00 €
Product Information
- Catalog #AvailabilityPrice
- Sensor cable S7/DIN, length 1 m
Product Information
- Catalog #AvailabilityPrice
- Sensor cable S7/Lemo, length 1 m
Product Information
- Catalog #PkAvailabilityPrice
- Electrolyte 1M KNO3, silver-free, 100 mlElectrolyte 1M KNO3, silver-free, 100 ml620-4934100 mlLog in for availability1 * 100 mL - 33,90 €



