pH electrodes, InLab®, Expert Series
About this item
Ideal for critical samples such as suspensions or samples of unknown composition (e.g. wastewater).
- ARGENTHAL™ reference system with silver-ion trap
- Solid XEROLYT® polymer electrolyte: No need for a junction (no possibility of contamination)
- Unbreakable PEEK shaft
- Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
pH range: 0 to 14
Temperature range: 0 to 100 °C
Temperature probe: NTC30 kOhm for InLab® Expert Pro and InLab® Expert NTC30; PT1000 for InLab® Expert DIN and InLab® Expert PT1000
Ordering information: Supplied with test certificate.
7 Options Available Below
Product Details & Documents
Ideal for critical samples such as suspensions or samples of unknown composition (e.g. wastewater).
- ARGENTHAL™ reference system with silver-ion trap
- Solid XEROLYT® polymer electrolyte: No need for a junction (no possibility of contamination)
- Unbreakable PEEK shaft
- Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
pH range: 0 to 14
Temperature range: 0 to 100 °C
Temperature probe: NTC30 kOhm for InLab® Expert Pro and InLab® Expert NTC30; PT1000 for InLab® Expert DIN and InLab® Expert PT1000
Ordering information: Supplied with test certificate.
Documents
Certificates
Search by Lot NumberMETT51343101_TDS_EN
Datasheet
Mettler InLab® Sensors
Product Literature
Product Family Options
Product Information
- TypepHTemp.MaterialElectrolyteØ×LConnectionForAvailabilityPrice
- 662-2842InLab® Expert DINInLab® Expert DINInLab® Expert DIN0 - 140...+100 °CPEEKXEROLYT® polymer12×120 mm1,2 m cable; DIN 19262/4 mmWastewater, highly saline solutions; gels, soaps and shampoos; proteinaceous media; beverages; suspended solids, oily samplesLog in for availability1 * 1 ST329,00 €
- 662-2843InLab® Expert NTC30InLab® Expert NTC30InLab® Expert NTC300 - 140...+100 °CPEEKXEROLYT® polymer12×120 mmMultiPin™Wastewater, highly saline solutions; gels, soaps and shampoos; proteinaceous media; beverages; suspended solids, oily samplesLog in for availability1 * 1 ST319,00 €
- 662-2844InLab® Expert PT1000InLab® Expert PT1000InLab® Expert PT10000 - 140...+100 °CPEEKXEROLYT® polymer12×120 mmMultiPin™Wastewater, highly saline solutions; gels, soaps and shampoos; proteinaceous media; beverages; suspended solids, oily samplesLog in for availability1 * 1 ST319,00 €
- 662-2840InLab® Expert ProInLab® Expert ProInLab® Expert Pro0 - 140...+100 °CPEEKXEROLYT® polymer12×120 mm1,2 m cable; BNC/RCA (Cinch)Wastewater, highly saline solutions; gels, soaps and shampoos; proteinaceous media; beverages; suspended solids, oily samplesLog in for availability364,00 €
- 662-2841InLab® Expert Pro-2m-ISMInLab® Expert Pro-2m-ISMInLab® Expert Pro-2m-ISM0 - 140...+100 °CPEEKXEROLYT® polymer12×120 mm2,0 m cable; BNC/RCA (Cinch)Wastewater, highly saline solutions; gels, soaps and shampoos; proteinaceous media; beverages; suspended solids, oily samplesLog in for availability1 * 1 ST431,00 €
- 662-2839InLab® ExpertInLab® ExpertInLab® Expert0 - 140...+100 °CPEEKXEROLYT® polymer12×120 mmS7Wastewater, highly saline solutions; gels, soaps and shampoos; proteinaceous media; beverages; suspended solids, oily samplesLog in for availability1 * 1 ST229,00 €
- 662-2738InLab® Expert GoInLab® Expert GoInLab® Expert Go0 - 140...+100 °CPEEKXEROLYT® polymer12×120 mmBNC/RCA (Cinch), 1,8 m cable (IP67)Portable applications in the lab and outdoorsLog in for availability411,00 €