pH and redox combination electrodes with temperature sensor, BlueLine
Catalog #
Jump to:
Product Details & Documents
The BlueLine family comprises three ranges of combination electrodes: Robust electrodes with gel electrolyte and plastic shaft for general use, liquid-electrolyte sensors for more critical measurements and special sensors.
- Special electrodes for redox measurements, surface measurements and food applications
- Gel electrolyte requires low maintenance and is not refillable
- ID sensors are equipped with a transponder, carrying the information of its type and serial number as well as the date and result of the last calibration
- IDS sensors automatically identify themselves with their serial numbers and type designation and transmit their calibration data when connected to a measuring device
pH range: 0 - 14
Accessories
Product Information
- Catalog #AvailabilityPrice
- Cable, 3 m, with S7/DIN plug
Product Information
- Catalog #AvailabilityPrice
- Cable, 1 m, with S7/BNC plug
Product Information
- Catalog #AvailabilityPrice
- Cable, 1 m, with S7/DIN plug