Orders LinkContactUsLinkComponent
pH Electrodes, SenTix®
Supplier:   WTW
pH Electrodes, SenTix®
Supplier:  WTW
Certificates
pH Electrodes, SenTix®
42 Options Available Below
Product Details & Documents
These proven and tested pH electrodes are easy to use and offer high quality and precision.
  • Electrodes with gel electrolyte and a plastic body are robust and shock-proof, the membrane glass is safeguarded by a protective shield
  • Electrodes with liquid electrolyte: Capillary-effect of the platinum wire guarantees constant outflow and prevents contamination by precipitates or even dirt
  • SenTix® HW and HWS electrodes: Electrolyte flow speed can be adjusted for optimal use
Optimised membrane glass guarantees quick measurement even at low temperatures. The easy to use fill-hole cover prevents electrolyte from leaking out during storage.
* with integrated temperature probe (NTC 30 KOhm).
Product Family Options
SenTix® electrodes (general applications: For use in the laboratory or for field work)
Product Information
  • Type
    pH
    Temp.
    Material
    Electrolyte
    Ø×L
    Connection
    For
    Availability
    Price
  • 662-1301
    pH electrode SenTix® 20
    pH electrode SenTix® 20
    SenTix® 20
    0 - 14
    0...+80 °C
    Plastic
    Gel
    12×120 mm
    S7**
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €
  • 662-1302
    pH electrode SenTix® 21
    pH electrode SenTix® 21
    SenTix® 21
    0 - 14
    0...+80 °C
    Plastic
    Gel
    12×120 mm
    DIN, fixed 1 m cable
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 191,00 €
    191,00 €
    1 * 1 ST
       191,00 €
  • 662-1303
    pH electrode SenTix® 21-3
    pH electrode SenTix® 21-3
    SenTix® 21-3
    0 - 14
    0...+80 °C
    Plastic
    Gel
    12×120 mm
    DIN, fixed 3 m cable
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 263,00 €
    263,00 €
    1 * 1 ST
       263,00 €
  • 662-1304
    pH electrode SenTix® 22
    pH electrode SenTix® 22
    SenTix® 22
    0 - 14
    0...+80 °C
    Plastic
    Gel
    12×120 mm
    BNC, fixed 1 m cable
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 206,00 €
    206,00 €
    1 * 1 ST
       206,00 €
  • 662-1305
    pH electrode SenTix® 41, with temperature sensor
    pH electrode SenTix® 41, with temperature sensor
    SenTix® 41*
    0 - 14
    0...+80 °C
    Plastic
    Gel
    12×120 mm
    DIN + banana plug (1×4 mm), fixed 1 m cable
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 289,00 €
    289,00 €
    1 * 1 ST
       289,00 €
  • 662-1306
    pH electrode SenTix® 41-3, with temperature sensor
    pH electrode SenTix® 41-3, with temperature sensor
    Sentix® 41-3M*
    0 - 14
    0...+80 °C
    Plastic
    Gel
    12×120 mm
    DIN + banana plug (1×4 mm), fixed 3 m cable
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 346,00 €
    346,00 €
    1 * 1 ST
       346,00 €
  • 662-1307
    pH electrode SenTix® 42, with temperature sensor
    pH electrode SenTix® 42, with temperature sensor
    Sentix® 42*
    0 - 14
    0...+80 °C
    Plastic
    Gel
    12×120 mm
    BNC + banana plug (1×4 mm), fixed 1 m cable
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 314,00 €
    314,00 €
    1 * 1 ST
       314,00 €
  • 662-1308
    pH electrode SenTix® 60
    pH electrode SenTix® 60
    SenTix® 60
    0 - 14
    0...+100 °C
    Glass
    KCl 3 mol/l, without Ag+
    12×122 mm
    S7**
    Laboratory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 213,00 €
    213,00 €
    1 * 1 ST
       213,00 €
  • 662-2151
    pH electrode SenTix® 61
    pH electrode SenTix® 61
    SenTix® 61
    0 - 14
    0...+100 °C
    Glass
    KCl 3 mol/l, without Ag+
    12×120 mm
    DIN, fixed 1 m cable
    Laboratory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,00 €
    223,00 €
    1 * 1 ST
       223,00 €
  • 662-1309
    pH electrode SenTix® 62
    pH electrode SenTix® 62
    SenTix® 62
    0 - 14
    0...+100 °C
    Glass
    KCl 3 mol/l, without Ag+
    12×120 mm
    BNC, fixed 1 m cable
    Laboratory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,00 €
    223,00 €
    1 * 1 ST
       223,00 €
  • 662-1315
    pH electrode SenTix® 81, with temperature sensor
    pH electrode SenTix® 81, with temperature sensor
    SenTix® 81*
    0 - 14
    0...+100 °C
    Glass
    KCl 3 mol/l, without Ag+
    12×120 mm
    DIN + banana plug (1×4 mm), fixed 1 m cable
    Laboratory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 339,00 €
    339,00 €
    1 * 1 ST
       339,00 €
  • 662-1316
    pH electrode SenTix® 82, with temperature sensor
    pH electrode SenTix® 82, with temperature sensor
    SenTix® 82*
    0 - 14
    0...+100 °C
    Glass
    KCl 3 mol/l, without Ag+
    12×120 mm
    BNC + banana plug (1×4 mm), fixed 1 m cable
    Laboratory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 345,00 €
    345,00 €
    1 * 1 ST
       345,00 €
  • 662-0106
    pH electrode SenTix® 51, with temperature sensor
    pH electrode SenTix® 51, with temperature sensor
    SenTix® 51*
    0 - 14
    0...+80 °C
    Plastic
    KCl 3 mol/l, without Ag+
    12×120 mm
    DIN + banana plug (1×4 mm), fixed 1 m cable
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 321,00 €
    321,00 €
    1 * 1 ST
       321,00 €
  • 662-1871
    pH electrode SenTix® 52, with temperature sensor
    pH electrode SenTix® 52, with temperature sensor
    SenTix® 52*
    0 - 14
    0...+80 °C
    Plastic
    KCl 3 mol/l, without Ag+
    12×120 mm
    BNC + banana plug (1×4 mm), fixed 1 m cable
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 346,00 €
    346,00 €
    1 * 1 ST
       346,00 €
  • 662-1277
    pH electrode SenTix® 91, with temperature sensor
    pH electrode SenTix® 91, with temperature sensor
    SenTix® 91*
    0 - 14
    0...+100 °C
    Glass
    KCl 3 mol/l, without Ag+
    12×170 mm
    DIN + banana plug (1×4 mm), fixed 1 m cable
    Laboratory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 576,00 €
    576,00 €
    1 * 1 ST
       576,00 €
  • WTWA103733
    SenTix® pH electrodes for penetration measurement
    SenTix® pH electrodes for penetration measurement
    SenTix® Sp-T
    2 - 13
    0 ... 80 °C
    Glass
    Referid®
    65/25 mm, 15/5 mm
    1 m fixed cable with water-proof DIN and 4 mm banana plug
    Laboratory/foods
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 503,00 €
    503,00 €
    1 * 1 ST
       503,00 €
  • WTWA103744
    SenTix® Top 940 IDS pH electrode with double reference system and polymer electrolyte
    SenTix® Top 940 IDS pH electrode with double reference system and polymer electrolyte
    SenTix® Top 940
    0 - 14
    –5…+100 °C
    Plastic
    Duralid®
    12×120 mm
    1,5 m fixed cable with waterproof digital plug
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • WTWA103792
    SenTix® Rx-T 900 IDS ORP electrode with temperature sensor
    SenTix® Rx-T 900 IDS ORP electrode with temperature sensor
    SenTix® Rx-T 900
    –5…+80 °C
    Plastic
    Gel
    12×120 mm
    1,5 m fixed cable with water-proof digital plug
    ORP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 299,00 €
    299,00 €
    1 * 1 ST
       299,00 €
  • WTWA103805
    pH electrode SenTix® 43, with temperature sensor
    pH electrode SenTix® 43, with temperature sensor
    Sentix® 43*
    0 - 14
    0...+80 °C
    Plastic
    Gel
    12×120 mm
    1 m fixed cable with water-proof DIN and 4 mm banana plug
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 332,00 €
    332,00 €
    1 * 1 ST
       332,00 €
  • WTWA103806
    pH electrode SenTix® 44, with temperature sensor
    pH electrode SenTix® 44, with temperature sensor
    Sentix® 44*
    0 - 14
    0...+80 °C
    Plastic
    Gel
    12×120 mm
    1 m fixed cable with BNC + banana plug
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 327,00 €
    327,00 €
    1 * 1 ST
       327,00 €
  • WTWA103807
    pH electrode SenTix® 46, with temperature sensor
    pH electrode SenTix® 46, with temperature sensor
    Sentix® 46*
    0 - 14
    0...+80 °C
    Plastic
    Gel
    12×120 mm
    1 m fixed cable with BNC plug and Cinch plug
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 332,00 €
    332,00 €
    1 * 1 ST
       332,00 €
  • WTWA103808
    pH electrode SenTix® 47, with temperature sensor
    pH electrode SenTix® 47, with temperature sensor
    Sentix® 47*
    0 - 14
    0...+80 °C
    Plastic
    Gel
    12×120 mm
    1 m fixed cable with BNC plug + 2.5 mm jack plug (for Sartorius devices)
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 334,00 €
    334,00 €
    1 * 1 ST
       334,00 €
  • WTWA103809
    pH electrode SenTix® 57, with temperature sensor
    pH electrode SenTix® 57, with temperature sensor
    Sentix® 57*
    0 - 14
    0...+80 °C
    Plastic
    KCl 3 mol/l Ag+ free
    12×120 mm
    1 m fixed cable with BNC plug and 2,5 mm jack plug
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 345,00 €
    345,00 €
    1 * 1 ST
       345,00 €
  • WTWA103810
    pH electrode SenTix® 83, with temperature sensor
    pH electrode SenTix® 83, with temperature sensor
    Sentix® 83*
    0 - 14
    0…+100 °C
    Glass
    KCl 3 mol/l Ag+ free
    12×120 mm
    1 m fixed cable with waterproof DIN plug + 4 mm banana plug
    Laboratory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 385,00 €
    385,00 €
    1 * 1 ST
       385,00 €
  • WTWA103811
    pH electrode SenTix® 84, with temperature sensor
    pH electrode SenTix® 84, with temperature sensor
    Sentix® 84*
    0 - 14
    0…+100 °C
    Glass
    KCl 3 mol/l Ag+ free
    12×120 mm
    1 m fixed cable with BNC and 4 mm banana plug
    Laboratory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 385,00 €
    385,00 €
    1 * 1 ST
       385,00 €
  • WTWA103812
    pH electrode SenTix® 85, with temperature sensor
    pH electrode SenTix® 85, with temperature sensor
    Sentix® 85*
    0 - 14
    0…+100 °C
    Glass
    KCl 3 mol/l Ag+ free
    12×120 mm
    1 m fixed cable with BNC + 2×4 mm banana plug
    Laboratory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 381,00 €
    381,00 €
    1 * 1 ST
       381,00 €
  • WTWA103813
    pH electrode SenTix® 86, with temperature sensor
    pH electrode SenTix® 86, with temperature sensor
    Sentix® 86*
    0 - 14
    0…+100 °C
    Glass
    KCl 3 mol/l Ag+ free
    12×120 mm
    1 m fixed cable with BNC plug and Cinch plug
    Laboratory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 371,00 €
    371,00 €
    1 * 1 ST
       371,00 €
  • WTWA103814
    pH electrode SenTix® 87, with temperature sensor
    pH electrode SenTix® 87, with temperature sensor
    Sentix® 87*
    0 - 14
    0…+100 °C
    Glass
    KCl 3 mol/l Ag+ free
    12×120 mm
    1 m fixed cable with BNC plug + 2,5 mm jack plug
    Laboratory
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 371,00 €
    371,00 €
    1 * 1 ST
       371,00 €
  • WTWA103815
    SenTix® Rx ORP electrode
    SenTix® Rx ORP electrode
    Sentix® Rx
    –5…+80 °C
    Plastic
    Gel
    12×120 mm
    S7 plug head, without cable
    ORP
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 248,00 €
    248,00 €
    1 * 1 ST
       248,00 €
  • WTWA103816
    SenTix® Top 41 pH electrodes
    SenTix® Top 41 pH electrodes
    SenTix® Top 41
    0 - 14
    –5…+100 °C
    Plastic
    Duralid®
    12×120 mm
    1 m fixed cable with waterproof DIN plug and 4 mm banana plug
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 339,00 €
    339,00 €
    1 * 1 ST
       339,00 €
  • WTWA103817
    SenTix® Top 46 pH electrodes
    SenTix® Top 46 pH electrodes
    SenTix® Top 46
    0 - 14
    –5…+100 °C
    Plastic
    Duralid®
    12×120 mm
    1 m fixed cable with BNC plug + cinch plug
    Field
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 364,00 €
    364,00 €
    1 * 1 ST
       364,00 €
SenTix® special electrodes
Product Information
  • Type
    pH
    Temp.
    Material
    Electrolyte
    Ø×L
    Connection
    For
    Availability
    Price
  • 662-1317
    pH electrode SenTix® H with ground joint diaphragm
    pH electrode SenTix® H with ground joint diaphragm
    SenTix® H
    0 - 14
    0...+80 °C
    Glass
    KCl 3 mol/l, without Ag+
    12×165 mm
    S7**
    Highly alkaline and highly saltine solutions
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 375,00 €
    375,00 €
    1 * 1 ST
       375,00 €
  • 662-0291
    pH electrode SenTix® SP
    pH electrode SenTix® SP
    SenTix® SP
    2 - 13
    0...+80 °C
    Glass
    Referid®
    15/5×65/25 mm
    S7**
    Penetration (bread, cheese, meat, fruit juice, solids)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 431,00 €
    431,00 €
    1 * 1 ST
       431,00 €
  • 662-1318
    pH electrode SenTix® Sur
    pH electrode SenTix® Sur
    SenTix® Sur
    2 - 13
    0...+50 °C
    Glass
    Referid®
    12×120 mm
    S7**
    Flat surfaces (leather, paper, skin, solids)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 385,00 €
    385,00 €
    1 * 1 ST
       385,00 €
  • 662-1319
    pH electrode SenTix® MIC
    pH electrode SenTix® MIC
    SenTix® MIC
    0 - 14
    0...+100 °C
    Glass
    KCl 3 mol/l, without Ag+
    12/5×40/80 mm
    S7**
    Small samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 430,00 €
    430,00 €
    1 * 1 ST
       430,00 €
  • 662-1400
    SenTix® ORP combination electrode
    SenTix® ORP combination electrode
    SenTix® ORP
    -
    0...+100 °C
    Glass
    KCl 3 mol/l
    12×120 mm
    S7**
    Wastewater, general purposes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 303,00 €
    303,00 €
    1 * 1 ST
       303,00 €
  • 662-1213
    pH electrode SenTix® HW (high concentration samples)
    pH electrode SenTix® HW (high concentration samples)
    SenTix® HW
    0 - 14
    0...+60 °C
    Glass
    KCl 3 mol/l, without Ag+
    12×170 mm
    S7**
    Solutions with low ion contents, emulsions, suspensions
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 355,00 €
    355,00 €
    1 * 1 ST
       355,00 €
  • 662-1275
    pH electrode SenTix® MIC-D (small samples)
    pH electrode SenTix® MIC-D (small samples)
    SenTix® MIC-D
    0 - 14
    –5...+100 °C
    Glass
    KCl 3 mol/L, Ag+ free
    3×96 mm
    DIN, fixed 1-m cable
    Small samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 501,00 €
    501,00 €
    1 * 1 ST
       501,00 €
  • 662-1872
    pH electrode SenTix® MIC-B
    pH electrode SenTix® MIC-B
    SenTix® MIC-B
    0 - 14
    –5...+100 °C
    Glass
    KCl 3 mol/l, without Ag+
    3×96 mm
    BNC, fixed 1-m cable
    Small samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 501,00 €
    501,00 €
    1 * 1 ST
       501,00 €
  • 662-2266
    SenTix® Ag combination electrode (special ORP electrode for argentometry)
    SenTix® Ag combination electrode (special ORP electrode for argentometry)
    SenTix® Ag
    -
    –5...+100 °C
    Glass
    2 mol/l KNO3 + 0,001 mol/l KCl
    12×120 mm
    S7**
    Laboratory/Argentometry
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 368,00 €
    368,00 €
    1 * 1 ST
       368,00 €
  • 662-1682
    pH electrode SenTix® HWD with temperature sensor and ground joint diaphragm
    pH electrode SenTix® HWD with temperature sensor and ground joint diaphragm
    SenTix® HWD*
    0 - 14
    –5...+100 °C
    Glass
    KCl 3 mol/L, Ag+ free
    12×170 mm
    DIN + banana plug (1×4 mm), fixed 1 m cable
    Solutions with low ion contents, emulsions, suspensions
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 499,00 €
    499,00 €
    1 * 1 ST
       499,00 €
  • 662-1999
    pH electrode SenTix® RJD with temperature sensor
    pH electrode SenTix® RJD with temperature sensor
    SenTix® RJD*
    2 - 13
    0...+80 °C
    Glass
    Polymer
    12×120 mm
    DIN + banana plug (1×4 mm), fixed 1 m cable
    Laboratory, polluted probes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 411,00 €
    411,00 €
    1 * 1 ST
       411,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cable, 3 m, with S7/DIN plug
    Cable, 3 m, with S7/DIN plug
    662-1326
    Log in for availability
    1 * 1 ST - 92,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Cable, 1 m, with S7/BNC plug
    Cable, 1 m, with S7/BNC plug
    662-1327
    Log in for availability
    1 * 1 ST - 66,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Cable, 1 m, with S7/DIN plug
    Cable, 1 m, with S7/DIN plug
    662-1425
    Log in for availability
    1 * 1 ST - 62,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Electrolyte with 2 mol/L KNO₃ and 0,001 mol/L KCl for combined ORP electrode with silver electrode
    Electrolyte with 2 mol/L KNO₃ and 0,001 mol/L KCl for combined ORP electrode with silver electrode
    WTWA109735
    Log in for availability
    1 * 250 mL - 59,90 €