Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Speciality pH Electrodes

VWR®, Speciality pH Electrodes

VWR®, Speciality pH Electrodes
Supplier:  VWR Collection
Certificates
VWR®, Speciality pH Electrodes
VWR®, Speciality pH Electrodes

17 Options Available Below

Product Details & Documents

pH electrodes with 1 m fixed cable or S7 connector.

  • Checked at five points during the manufacturing process and lot stamped to ensure quality
  • Storage soaker bottle to protect the electrode, keeping the item in top condition and instantly ready for use

pH range: 0 to 14

Temperature range: 0 to +100 °C (10 to 100 °C for flat tips)

Response time: 95% <1 second (95% <5 seconds for flat tips)

Product Family Options

With 1 m fixed cable

Product Information

  • Type
    Temp.
    Material
    Electrolyte
    Ø×L
    Connection
    Length
    For
    Availability
    Price
  • Thin, ceramic junction
    0...100 °C
    Glass
    3,5 M KCl
    9,5×300 mm
    BNC
    Tap water, seawater, desinfectants, soap
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 183,00 €
    183,00 €
    1 * 1 ST
       183,00 €

  • Thin, Pellon junction
    0...100 °C
    Epoxy
    Gel
    9×300 mm
    BNC
    Tap water, seawater, desinfectants, soap
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 165,00 €
    165,00 €
    1 * 1 ST
       165,00 €

  • Thin, Pellon junction
    0...100 °C
    Epoxy
    Gel
    9×300 mm
    DIN
    Tap water, seawater, desinfectants, soap
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 199,00 €
    199,00 €
    1 * 1 ST
       199,00 €

  • Semi-micro, Pellon junction
    0...100 °C
    Epoxy
    Gel
    6×150 mm
    BNC
    Tap water, seawater, desinfectants, soap
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 195,00 €
    195,00 €
    1 * 1 ST
       195,00 €

  • Semi-micro, Pellon junction
    0...100 °C
    Epoxy
    Gel
    6×150 mm
    DIN
    Tap water,seawater, desinfectants, soap
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 213,00 €
    213,00 €
    1 * 1 ST
       213,00 €

  • Large surface flat tip, HDPE junction
    0...100 °C
    Epoxy
    Gel
    15×115 mm
    BNC
    Agar, food, cosmetics, leather, water, viscous samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 171,00 €
    171,00 €
    1 * 1 ST
       171,00 €

  • Large surface flat tip, HDPE junction
    0...100 °C
    Epoxy
    Gel
    15×115 mm
    DIN
    Agar, food, cosmetics, leather, water, viscous samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 207,00 €
    207,00 €
    1 * 1 ST
       207,00 €

  • Flat tip, double junction (HDPE/Pellon)
    10...100 °C
    Epoxy
    Gel
    12×150 mm
    BNC
    Agar, food and beverage, serum, soil, Tris buffers, viscous samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,00 €
    223,00 €
    1 * 1 ST
       223,00 €

  • Flat tip, double junction (HDPE/Pellon)
    10...100 °C
    Glass
    3,5 M KCl
    12×150 mm
    BNC
    Agar, food and beverage, serum, soil, Tris buffers, viscous samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 235,00 €
    235,00 €
    1 * 1 ST
       235,00 €

  • Flat tip, double junction (HDPE/Pellon)
    0...100 °C
    Epoxy
    Gel
    12×150 mm
    DIN
    Agar, food and beverage, serum, soil, Tris buffers, viscous samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 268,00 €
    268,00 €
    1 * 1 ST
       268,00 €

  • Flat tip, double junction (HDPE/Pellon), fluoride resistant bulb
    0...100 °C
    Epoxy
    3,5 M KCl/sat. KNO3
    12×150 mm
    DIN
    Agar plates, electrophoresis gels
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €

  • Spear tip, double junction (PVDF/Pellon)
    0...100 °C
    Epoxy
    Gel
    12/9×150 mm
    BNC
    Bread, butter, cheese, fats, meat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 191,00 €
    191,00 €
    1 * 1 ST
       191,00 €

  • Spear tip, double junction (PVDF/Pellon)
    0...100 °C
    Epoxy
    Gel
    12/9×150 mm
    DIN
    Bread, butter, cheese, fats, meat
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 213,00 €
    213,00 €
    1 * 1 ST
       213,00 €

With S7 connector

Product Information

  • Type
    Temp.
    Material
    Electrolyte
    Ø×L
    Connection
    Length
    For
    Availability
    Price
  • Thin, Pellon junction
    0...100 °C
    Epoxy
    Gel
    9×300 mm
    S7
    300 mm
    Distilled water, ultra-pure water, toothpaste, Tris Buffers, microbiology
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 199,00 €
    199,00 €
    1 * 1 ST
       199,00 €

  • Semi-micro, Pellon junction
    0...100 °C
    Epoxy
    Gel
    6×150 mm
    S7
    Distilled water, ultra-pure water, toothpaste, Tris buffers, microbiology
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 213,00 €
    213,00 €
    1 * 1 ST
       213,00 €

  • Flat tip, double junction (HDPE/Pellon)
    0...100 °C
    Epoxy
    Gel
    12×150 mm
    S7
    Agar, food and beverage, serum, soil, Tris buffers, viscous samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 268,00 €
    268,00 €
    1 * 1 ST
       268,00 €

  • Flat tip, double junction (HDPE/Pellon)
    0...100 °C
    Glass
    Gel
    12×151 mm
    S7
    Agar, food and beverage, serum, soil, Tris buffers, viscous samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 275,00 €
    275,00 €
    1 * 1 ST
       275,00 €

Low flow cells for pH measurement

Product Information

  • Catalog #
    Description
    Availability
    Price
  • Low flow cell 200 µl, acrylic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 411,00 €
    411,00 €
    1 * 1 ST
       411,00 €

Accessories

Product Information

  • Catalog #
    Length
    Availability
    Price
  • BNC connector cable, coaxial, Coax53
    BNC connector cable, coaxial, Coax53
    662-1410
    1 m
    Log in for availability
    1 * 1 ST - 54,90 €

Product Information

  • Catalog #
    Length
    Availability
    Price
  • DIN connector cable, coaxial, Coax54
    DIN connector cable, coaxial, Coax54
    662-1411
    1 m
    Log in for availability
    1 * 1 ST - 67,40 €