Order Entry
Belgium
Orders LinkContactUsLinkComponent
Wipes for delicate tasks, Multisoft

Wipes for delicate tasks, Multisoft

Wipes for delicate tasks, Multisoft
Certificates
Wipes for delicate tasks, Multisoft
Wipes for delicate tasks, Multisoft

2 Options Available Below

Product Details & Documents

Soft non woven cloths made of Airlaid material, ideal for sensitive applications.

  • Solvent resistant and highly absorbent
  • Tear resistant, voluminous and lint-free
  • Packed in convenient dispensing box for single sheet dispensing
Product Family Options

Product Information

  • Colour
    L×W
    Packed
    Availability
    Price
  • White
    340×320 mm
    Pack of 25
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 3,50 €
    3,50 €
    1 * 25 ST
       3,50 €

  • White
    340×320 mm
    20 packs of 25
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 93,90 €
    93,90 €
    1 * 500 ST
       93,90 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.