Orders LinkContactUsLinkComponent
Joseph paper
Joseph paper
Supplier:   PRAT DUMAS
Joseph paper
Supplier:  PRAT DUMAS
Certificates
Joseph paper
3 Options Available Below
Product Details & Documents
Joseph paper is ideal for precise cleaning applications. Superior quality for wiping or packaging delicate objects.
  • Low linting
  • Absorbs liquids easily and quickly
L×W (mm): 350×500
Weight: 30 g/m²
Product Family Options
Product Information
  • L×W
    Availability
    Price
  • 291-0014
    800 folded sheets
    800 folded sheets
    350×500 mm
    Log in for availability
    Log in for availability
    1 * 800 ST
    1 * 800 ST - 187,00 €
    187,00 €
    1 * 800 ST
       187,00 €
  • 291-0015
    40 folded sheets
    40 folded sheets
    350×500 mm
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 35,30 €
    35,30 €
    1 * 40 ST
       35,30 €
  • PRATD500407
    500 folded sheets
    500 folded sheets
    350×500 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 124,00 €
    124,00 €
    1 * 500 ST
       124,00 €