Watch glasses
Supplier: SciLabware
Jump to:
About this item
PYREX®, borosilicate glass, with fused edges.
- Excellent chemical resistance
- High temperature resistance
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.
8 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, with fused edges.
- Excellent chemical resistance
- High temperature resistance
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.
Product Family Options
Product Information
- ØAvailabilityPrice
- 216-2770Watch glass, PYREX® borosilicate glass with firepolished edgesWatch glass, PYREX® borosilicate glass with firepolished edges40 mmLog in for availability1 * 1 ST13,40 €Specifications:
More
Specifications
Cat. No.216-2770 - 216-2748Watch glass, PYREX® borosilicate glass with firepolished edgesWatch glass, PYREX® borosilicate glass with firepolished edges50 mmLog in for availability1 * 1 ST11,70 €Specifications:
More
Specifications
Cat. No.216-2748 - BIBB1060/09DWatch glass, PYREX® borosilicate glassSpecifications:
More
Specifications
Cat. No.BIBB1060/09D - BIBB1060/10DWatch glass, PYREX® borosilicate glassSpecifications:
More
Specifications
Cat. No.BIBB1060/10D - 216-2751Watch glass, PYREX® borosilicate glass with firepolished edgesWatch glass, PYREX® borosilicate glass with firepolished edges100 mmLog in for availability1 * 10 ST159,00 €Specifications:
More
Specifications
Cat. No.216-2751 - 216-2752Watch glass, PYREX® borosilicate glass with firepolished edgesWatch glass, PYREX® borosilicate glass with firepolished edges125 mmLog in for availability1 * 1 ST22,80 €Specifications:
More
Specifications
Cat. No.216-2752 - 216-2777Watch glass, PYREX® borosilicate glass with firepolished edgesWatch glass, PYREX® borosilicate glass with firepolished edges150 mmLog in for availability1 * 1 ST27,80 €Specifications:
More
Specifications
Cat. No.216-2777 - 216-2753Watch glass, PYREX® borosilicate glass with firepolished edgesWatch glass, PYREX® borosilicate glass with firepolished edges200 mmLog in for availability1 * 10 ST340,00 €Specifications:
More
Specifications
Cat. No.216-2753