Ubbelohde viscometers, DIN
Supplier: SI Analytics
Certificates
About this item
Ubbelohde viscometers with suspended ball level for determination of relative or absolute kinematic viscosity.
- Additional filling and cleaning tube
- Glass threads ensure safe operational use
- Calibrated with constant for automatic measurement
Certifications: DIN 51562-1, ISO 3105, BS 133, NFT 60-100
Specifications
- Catalog No:
- 616-3380
- 616-3385
- 616-3384
- 616-3387
- 616-3383
- 616-3382
- Length:
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm
- 290 mm