Orders LinkContactUsLinkComponent
Scintillation vials, WHEATON®
Scintillation vials, WHEATON®
Supplier:   DWK Life Sciences
Scintillation vials, WHEATON®
Supplier:  DWK Life Sciences
Certificates
Scintillation vials, WHEATON®
Scintillation vials, WHEATON®
14 Options Available Below
Product Details & Documents
These scintillation vials made from HDPE are available with screw cap attached or separately in PP or PE and different liner material depending on the application.
Product Family Options
Product Information
  • Capacity
    Ø×H
    Cap included
    Cap type
    Cap Size
    Cap material
    Septum
    Availability
    Price
  • 20,00 ml
    27×61 mm
    Yes
    Attached
    22 mm
    PP
    Foamed PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 263,00 €
    263,00 €
    1 * 500 ST
       263,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Attached
    22 mm
    PP
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 263,00 €
    263,00 €
    1 * 500 ST
       263,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Attached
    22 mm
    Urea
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 312,00 €
    312,00 €
    1 * 500 ST
       312,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Attached
    22 mm
    PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 341,00 €
    341,00 €
    1 * 500 ST
       341,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Attached
    22 mm
    Urea
    PE cone
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 425,00 €
    425,00 €
    1 * 500 ST
       425,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Separate
    22 mm
    PP
    Foamed PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 264,00 €
    264,00 €
    1 * 500 ST
       264,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Separate
    22 mm
    PP
    Foamed PE
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 419,00 €
    419,00 €
    1 * 1.000 ST
       419,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Separate
    22 mm
    PP
    Metal foil
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 267,00 €
    267,00 €
    1 * 500 ST
       267,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Separate
    22 mm
    PP
    Metal foil
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 419,00 €
    419,00 €
    1 * 1.000 ST
       419,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Separate
    22 mm
    PE
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 332,00 €
    332,00 €
    1 * 500 ST
       332,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Separate
    22 mm
    PE
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 550,00 €
    550,00 €
    1 * 1.000 ST
       550,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Separate
    22 mm
    Urea
    Metal foil
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 516,00 €
    516,00 €
    1 * 1.000 ST
       516,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Separate
    22 mm
    Urea
    PE cone
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 425,00 €
    425,00 €
    1 * 500 ST
       425,00 €
  • 20,00 ml
    27×61 mm
    Yes
    Separate
    22 mm
    Urea
    PE cone
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 718,00 €
    718,00 €
    1 * 1.000 ST
       718,00 €