Orders LinkContactUsLinkComponent
Scintillation vials
Scintillation vials
Supplier:   KARTELL
Scintillation vials
Supplier:  KARTELL
Certificates
Scintillation vials
Scintillation vials
2 Options Available Below
Product Details & Documents
HDPE, translucent. Compatible with all scintillation counters. A special screw cap ensures leakproof handling.
Documents
Product Family Options
Product Information
  • Capacity
    Ø×H
    Cap included
    Availability
    Price
  • 4,00 ml
    13,7×53,1 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 289,00 €
    289,00 €
    1 * 2.000 ST
       289,00 €
  • 20,00 ml
    26,5×58,5 mm
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 322,00 €
    322,00 €
    1 * 1.000 ST
       322,00 €