Sample vials, trident vials, squat
Supplier: MURRAY S
Jump to:
About this item
Vials made from neutral glass with snap-on lid. Designed for a wide variety of uses in the laboratory, production, more.
- Wide opening for easy filling and emptying
- With beaded rim and sealing snap-on lid
- Robust and economical, will withstand moderate temperature change
Packaging: For production related reasons, the number of vials per box may vary.
4 Options Available Below
Product Details & Documents
Vials made from neutral glass with snap-on lid. Designed for a wide variety of uses in the laboratory, production, more.
- Wide opening for easy filling and emptying
- With beaded rim and sealing snap-on lid
- Robust and economical, will withstand moderate temperature change
Packaging: For production related reasons, the number of vials per box may vary.
Product Family Options
Product Information