Orders LinkContactUsLinkComponent
VWR® ND9, Short Thread Vials
VWR® ND9, Short Thread Vials
Supplier:  LA PHA PACK
Certificates
VWR® ND9, Short Thread Vials
VWR® ND9, Short Thread Vials
5 Options Available Below
Product Details & Documents
Standard glass or plastic vials for GC and HPLC, micro-inserts, closures and kits. Can be used universally on almost all autosamplers, preferred type of vial for Waters and Agilent.
  • Wide opening enables easy filling
  • Closures with different types of septa can be supplied with differently coloured caps; blue cap also available with closed top
  • Pre-cut closures for HPLC needles are available, where only the silicone is slitted, but the PTFE remains intact (no concentration changes; no contamination due to impurities extracted from the silicone)
  • Synthetic material often used in Agilent red rubber/PTFE septa has been added to the product line (this is a softer material with less fragmentation than natural rubber/TEF)
Special short thread cap for LC-MS and GC-MS applications: One component closure, no bleeding, absolutely inert, tight, pierceable and chemical resistant like PTFE.
The use of HPLC and GC certified vial kits are an important way of reducing sources of error and ensuring that processes are reproducible.
 : Kits include: 1,5 ml short thread vial (11,6×32 mm), short thread cap PP blue, red or transparent with 6 mm central hole and septum.
Product Family Options
Short thread vials, ND9; 1000-Pack
Product Information
  • Capacity
    Ø×H
    Colour
    Cap included
    Availability
    Price
  • Vials
    1,50 ml
    11,6×32 mm
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 24,80 €
    24,80 €
    1 * 100 ST
       24,80 €
  • 548-3029
    Vials, with label
    Vials, with label
    1,50 ml
    11,6×32 mm
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 23,90 €
    23,90 €
    1 * 100 ST
       23,90 €
  • 548-3030
    Vials, with label
    Vials, with label
    1,50 ml
    11,6×32 mm
    Amber
    No
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 27,70 €
    27,70 €
    1 * 100 ST
       27,70 €
Short thread vials in plastic, ND9
Product Information
  • Capacity
    Ø×H
    Colour
    Cap included
    Availability
    Price
  • 548-3094
    Vials, conical insert
    Vials, conical insert
    0,30 ml
    11,6×32 mm
    Transparent
    No
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 29,70 €
    29,70 €
    1 * 100 ST
       29,70 €
  • LAPH11191205
    Vials, with filling lines
    Vials, with filling lines
    1,50 ml
    11,6×32 mm
    Transparent
    No
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 305,00 €
    305,00 €
    1 * 1.000 ST
       305,00 €
Micro-inserts for wide opening
Product Information
  • Catalog #
    Colour
    Ø×H
    Capacity
    Cap included
    Description
    Availability
    Price
  • Clear
    6×31 mm
    0,10 ml
    No
    Micro-insert, 15 mm top
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 32,80 €
    32,80 €
    1 * 100 ST
       32,80 €
Short screw caps with central hole, 6 mm and septa, ND9; 1000-Pack
Product Information
  • Catalog #
    Hardness
    Septum
    Thickness
    Type
    Availability
    Price
  • 60° shore A
    Natural rubber red-orange/TEF transparent
    1,0 mm
    PP transparent
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 24,00 €
    24,00 €
    1 * 100 ST
       24,00 €
Product Information
  • Catalog #
    Hardness
    Septum
    Thickness
    Type
    Availability
    Price
  • 45° shore A
    PTFE red/silicone white/PTFE red
    1,0 mm
    PP transparent
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 47,60 €
    47,60 €
    1 * 100 ST
       47,60 €
Product Information
  • Catalog #
    Hardness
    Septum
    Thickness
    Type
    Availability
    Price
  • 45° shore A
    Red rubber/PTFE beige
    1,0 mm
    PP blue
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 283,00 €
    283,00 €
    1 * 1.000 ST
       283,00 €
Product Information
  • Catalog #
    Hardness
    Septum
    Thickness
    Type
    Availability
    Price
  • 55° shore A
    Silicone white/PTFE red
    1,0 mm
    PP blue
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 32,00 €
    32,00 €
    1 * 100 ST
       32,00 €
Product Information
  • Catalog #
    Hardness
    Septum
    Thickness
    Type
    Availability
    Price
  • 55° shore A
    Silicone white/PTFE red
    1,0 mm
    PP transparent
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 36,20 €
    36,20 €
    1 * 100 ST
       36,20 €