Headspace crimp caps with 20 mm septa
Supplier: Agilent
Certificates
About this item
Frequently Bought Together
Certified headspace crimp top vials
Supplier: Agilent
Diethylamine for synthesis
Supplier: Merck
Glass crimp top headspace vials, 20 ml, level 2 high-throughput applications, SureSTART™
Supplier: Thermo Fisher Scientific
NEEDLE C (20/51/3) 1 * 6 items
Supplier: HAMILTON BONADUZ
Calcium chloride 1.0 M CaCl2 ≥98% volumetric solution, Fluka™
Supplier: Honeywell Chemicals
1-Butanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Acetone ≥99%
Supplier: Thermo Fisher Scientific
Capillary column ferrules for gas chromatography
Supplier: Agilent
SYRINGE 100R-GT 100UL CALIBRATED 1 * 1 items
Supplier: Trajan Scientific and Medical
Fused silica tubing
Supplier: Agilent
ThinCert® 96-Well HTS Inserts
Supplier: Greiner Bio-One
Chlorobenzene, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Fraction collector supplies
Supplier: Agilent
Headspace crimp top vials, CrossLab
Supplier: Agilent
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Jerricans, Narrow Neck, HDPE, UN-X Approved
Supplier: RIXIUS