Orders LinkContactUsLinkComponent
Non-return valve
Non-return valve
7 mm ID
Catalog # 229-3591
Supplier:   Lenz Laborglas
Non-return valve
Catalog # 229-3591
Supplier:  Lenz Laborglas
Non-return valve
Non-return valve
7 mm ID
Price
25,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, clear. Prevent pressure loss or backflow. Simple design, cannot be dismantled.
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • With tapered ends to ensure that the tube is held firmly
Documents
Specifications
  • Tubing size:
    7 mm ID